Merck
  • Enhancement of the proapoptotic properties of newcastle disease virus promotes tumor remission in syngeneic murine cancer models.
Enhancement of the proapoptotic properties of newcastle disease virus promotes tumor remission in syngeneic murine cancer models.
Molecular cancer therapeutics (2015-03-13)
Sara Cuadrado-Castano, Juan Ayllon, Mena Mansour, Janis de la Iglesia-Vicente, Stefan Jordan, Shashank Tripathi, Adolfo García-Sastre, Enrique Villar
ABSTRACT
Newcastle disease virus (NDV) is considered a promising agent for cancer therapy due to its oncolytic properties. These include preferential replication in transformed cells, induction of innate and adaptive immune responses within tumors, and cytopathic effects in infected tumor cells due to the activation of apoptosis. To enhance the latter and thus possibly enhance the overall oncolytic activity of NDV, we generated a recombinant NDV encoding the human TNF receptor Fas (rNDV-B1/Fas). rNDV-B1/Fas replicates to similar titers as its wild-type (rNDV-B1) counterpart; however, overexpression of Fas in infected cells leads to higher levels of cytotoxicity correlated with faster and increased apoptosis responses, in which both the intrinsic and extrinsic pathways are activated earlier. Furthermore, in vivo studies in syngeneic murine melanoma models show an enhancement of the oncolytic properties of rNDV-B1/Fas, with major improvements in survival and tumor remission. Altogether, our data suggest that upregulation of the proapoptotic function of NDV is a viable approach to enhance its antitumor properties and adds to the currently known, rationally based strategies to design optimized therapeutic viral vectors for the treatment of cancer.
Product Description
SAFC
L-Glutamine
Sigma-Aldrich
L-Glutamine
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
MISSION® esiRNA, targeting human FAS
Sigma-Aldrich
Caspase-8 Inhibitor II, InSolution