Merck
  • Effects of malted and non-malted whole-grain wheat on metabolic and inflammatory biomarkers in overweight/obese adults: a randomised crossover pilot study.
Effects of malted and non-malted whole-grain wheat on metabolic and inflammatory biomarkers in overweight/obese adults: a randomised crossover pilot study.
Food chemistry (2015-10-17)
Kristina Nelson, Michael L Mathai, John F Ashton, Osaana N Donkor, Todor Vasiljevic, Ravikumar Mamilla, Lily Stojanovska
ABSTRACT
Metabolic dysfunction in obesity may be attenuated by whole-grain intake, which has been attributed to synergistic actions of bioactive compounds. Germination/malting may increase grain bioactives, including polyphenols, however biological effects compared with non-germinated grains are unclear. Polyphenols and biological effects were compared between malted-wheat (MLT) and whole-grain wheat (CON) breakfast cereals. Polyphenol content and antioxidant activity were significantly higher (P<0.01 and P<0.05, respectively) in MLT. Corresponding obesity-related biomarkers were measured in 10 overweight/obese adults in a 2×4-week double-blind, randomised, crossover trial. Following both interventions, diastolic blood pressure reduced significantly with time (P<0.05) and low-density lipoprotein increased slightly (P<0.05) over time. A significant time*cereal effect (P<0.05) was found for insulin resistance, decreasing following CON and increasing with MLT. No other significant metabolic or inflammatory differences were found. Whilst MLT contained significantly increased polyphenols, cumulative effects in attenuating obesity-related metabolic dysfunction may require increased consumption.
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Phosphoric acid, ACS reagent, ≥85 wt. % in H2O
Sigma-Aldrich
Ultrapure Acetonitrile
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Phosphoric acid, ACS reagent, ≥85 wt. % in H2O
SAFC
Cholesterol, Plant-Derived, SyntheChol®
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Phosphoric acid-16O4 solution, 70 wt. % in D2O, 99.9 atom % 16O
Sigma-Aldrich
(−)-Epigallocatechin gallate, ≥95%
Sigma-Aldrich
(−)-Epigallocatechin gallate, ≥80% (HPLC), from green tea
Sigma-Aldrich
Phosphoric acid, BioReagent, suitable for insect cell culture, 85%
Sigma-Aldrich
Phosphoric acid, BioUltra, ≥85% (T)
Sigma-Aldrich
trans-Ferulic acid, 99%
Sigma-Aldrich
Phosphoric acid, 85 wt. % in H2O, 99.99% trace metals basis
Sigma-Aldrich
Sodium carbonate-12C, 99.9 atom % 12C
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Phosphoric acid, ≥85 wt. % in H2O, ≥99.999% trace metals basis
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Gallic acid, 97.5-102.5% (titration)
Sigma-Aldrich
Phosphoric acid, crystalline, ≥99.999% trace metals basis
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, for molecular biology
Sigma-Aldrich
Luteolin, ≥98% (TLC), powder
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
trans-Ferulic acid, ≥99%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis