Merck
  • The prognostic significance and relationship with body composition of CCR7-positive cells in colorectal cancer.
The prognostic significance and relationship with body composition of CCR7-positive cells in colorectal cancer.
Journal of surgical oncology (2015-07-17)
George Malietzis, Gui Han Lee, David Bernardo, Alexandra I F Blakemore, Stella C Knight, Morgan Moorghen, Hafid O Al-Hassi, John T Jenkins
ABSTRACT
The host local immune response (LIR) to cancer is a determinant of cancer outcome. Regulation of this local response is largely achieved through chemokine synthesis from the tumor microenvironment such as C-Chemokine-Receptor-7 (CCR7). We examined the LIR measured as CCR7 expression, in colorectal cancers (CRC) and explored relationships with body composition (BC) and survival. A study of paraffin-embedded tissue specimens was carried out in 116 patients with non-metastatic CRC. CCR7 expression was determined by immunohistochemistry. Analysis of computer tomography scans was used to calculate BC parameters. Survival analyses and multivariate regression models were used. High CCR7(+) cell density within the tumor stroma and at the margin was significantly associated with increased age, the presence of lymphovascular invasion, higher tumor stage, lymph node metastasis, high Klintrup-Makinen immune score, and myosteatosis. High CCR7(+) cell density in the tumor margin was significantly associated with shorter disease-free (DFS) and overall survival (OS) (P < 0.001). This was also significantly associated with shorter survival in multivariate analysis (HR = 8.87; 95%CI [2.51-31.3]; P < 0.01 for OS and HR = 4.72; 95%CI (1.24-12.9); P = 0.02 for DFS). Our results suggest that a specific immune microenvironment may be associated with altered host's BC and tumor behavior, and that CCR7 may serve as a novel prognostic biomarker.
Sigma-Aldrich
MISSION® esiRNA, targeting human CCR7
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture