Merck
  • Tamoxifen Inhibits TGF-β-Mediated Activation of Myofibroblasts by Blocking Non-Smad Signaling Through ERK1/2.
Tamoxifen Inhibits TGF-β-Mediated Activation of Myofibroblasts by Blocking Non-Smad Signaling Through ERK1/2.
Journal of cellular physiology (2015-06-23)
Jon M Carthy, Anders Sundqvist, Angelos Heldin, Hans van Dam, Dimitris Kletsas, Carl-Henrik Heldin, Aristidis Moustakas
ABSTRACT
Transforming growth factor-β (TGF-β) is a multifunctional cytokine which stimulates the differentiation of fibroblasts into myofibroblasts. Myofibroblasts are critical for normal wound healing, but also accumulate pathologically in a number of chronic inflammatory conditions where they are key contributors to aberrant tissue remodeling and fibrosis, and in cancer stroma. In the current study, we identified a role for tamoxifen as a potent inhibitor of the TGF-β-mediated activation of primary human skin and breast fibroblasts. Our data indicate that tamoxifen does not interfere with canonical Smad signaling downstream of TGF-β but rather blocks non-Smad signaling through ERK1/2 MAP-kinase and the AP-1 transcription factor FRA2. We further demonstrate by siRNA-mediated knockdown that FRA2 is critical for the induced expression of myogenic proteins in response to TGF-β. Functionally, TGF-β-stimulated fibroblast-mediated contraction of collagen gels was impaired in the presence of tamoxifen. Altogether, these data demonstrate that tamoxifen prevents myofibroblast differentiation and, therefore, may provide therapeutic benefits to patients suffering from chronic inflammatory conditions or cancer.
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Thrombopoietin human, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture, ≥98% (SDS-PAGE and HPLC)
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
2-Butanone, FCC, FG
Sigma-Aldrich
Anti-Fibronectin antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Thrombopoietin from mouse, recombinant, expressed in NSO cells, lyophilized powder, suitable for cell culture, >97% (SDS-PAGE)
Sigma-Aldrich
MISSION® esiRNA, targeting human FOSL2
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Formaldehyde solution, Molecular Biology, 36.5-38% in H2O
Sigma-Aldrich
Formaldehyde solution, Molecular Biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C