Merck
  • The effect of magnesium on partial sulphate removal from mine water as gypsum.
The effect of magnesium on partial sulphate removal from mine water as gypsum.
Journal of environmental management (2015-06-13)
Emma-Tuulia Tolonen, Jaakko Rämö, Ulla Lassi
ABSTRACT
The aim of this research was to investigate the effect of magnesium on the removal efficiency of sulphate as gypsum from mine water. The precipitation conditions were simulated with MINEQL + software and the simulation results were compared with the results from laboratory jar test experiments. Both the simulation and the laboratory results showed that magnesium in the mine water was maintaining sulphate in a soluble form as magnesium sulphate (MgSO4) at pH 9.6. Thus magnesium was preventing the removal of sulphate as gypsum (CaSO4·2H2O). However, change in the lime precipitation pH from 9.6 to 12.5 resulted in magnesium hydroxide (Mg(OH)2) precipitation and improved sulphate removal. Additionally, magnesium hydroxide could act as seed crystals for gypsum precipitation or co-precipitate sulphate further enhancing the removal of sulphate from mine water.
Sigma-Aldrich
Calcium oxide, 99.995% trace metals basis
Sigma-Aldrich
Calcium oxide, nanopowder, <160 nm particle size (BET), 98%
Sigma-Aldrich
Calcium oxide, ReagentPlus®, 99.9% trace metals basis
Sigma-Aldrich
Calcium oxide, anhydrous, powder, ≥99.99% trace metals basis