Merck
  • In Vitro and In Vivo Efficacy of Self-Assembling RGD Peptide Amphiphiles for Targeted Delivery of Paclitaxel.
In Vitro and In Vivo Efficacy of Self-Assembling RGD Peptide Amphiphiles for Targeted Delivery of Paclitaxel.
Pharmaceutical research (2015-06-13)
Poonam Saraf, Xiaoling Li, Lisa Wrischnik, Bhaskara Jasti
ABSTRACT
The objective of this work was to compare the efficacy of self-assembling cyclic and linear RGD peptide amphiphiles as carriers for delivering paclitaxel to αvβ3 integrin overexpressing tumors. Linear (C18-ADA5-RGD) and cyclic (C18-ADA5-cRGDfK) peptide amphiphiles were synthesized and characterized for CMC, aggregation number and micelle stability using fluorescence spectroscopy methods. Size and morphology of micelles was studied using TEM. Fluorescence polarization and confocal microscopy assays were established to compare binding and internalization of micelles. The targeting efficacy was studied in A2058 cells using cytotoxicity assay as well as in vivo in melanoma xenograft mouse model. The linear and cyclic RGD amphiphiles exhibited CMC of 25 and 8 μM, respectively, formed nano-sized spherical micelles and showed competitive binding to αvβ3 integrin protein. FITC-loaded RGD micelles rapidly internalized into A2058 melanoma cells. Paclitaxel-loaded RGD micelles exhibited higher cytotoxicity compared with free drug in A2058 cells in vitro as well as in vivo. Cyclic RGD micelles exhibited better targeting efficacy but were less effective compared to linear RGD micelles as drug delivery vehicle due to lower drug solubilization capacity and lesser kinetic stability. Results from the study proved the effectiveness of self-assembling low molecular weight RGD amphiphiles as carriers for targeted delivery of paclitaxel.
Sigma-Aldrich
Piperidine, ≥99.5%, purified by redistillation
Sigma-Aldrich
1,1′-Dioctadecyl-3,3,3′,3′-tetramethylindocarbocyanine perchlorate, BioReagent, suitable for fluorescence, ≥98.0% (TLC)
Sigma-Aldrich
Diethyl ether
Sigma-Aldrich
1,1′-Dioctadecyl-3,3,3′,3′-tetramethylindocarbocyanine perchlorate, 97%
Sigma-Aldrich
HATU, ≥98.0% (CHN)
Sigma-Aldrich
Fmoc-Gly-OH, ≥98.0% (T)
Sigma-Aldrich
HATU, 97%
Sigma-Aldrich
Piperidine, ReagentPlus®, 99%
Sigma-Aldrich
3,3′-Dioctadecyloxacarbocyanine perchlorate
Sigma-Aldrich
N,N-Dimethylformamide, for molecular biology, ≥99%
Sigma-Aldrich
N,N-Dimethylformamide, anhydrous, 99.8%
Sigma-Aldrich
Diethyl ether, contains 1 ppm BHT as inhibitor, anhydrous, ≥99.7%
SAFC
L-Glutamine
Sigma-Aldrich
ADA, ≥98% (titration)
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Triisopropylsilane, 98%
Sigma-Aldrich
Stearic acid, ≥95%, FCC, FG
Sigma-Aldrich
N,N-Diisopropylethylamine, purified by redistillation, 99.5%
Sigma-Aldrich
N,N-Diisopropylethylamine, ReagentPlus®, ≥99%
Sigma-Aldrich
2-Chlorotrityl chloride, ≥97.0% (AT)
Sigma-Aldrich
Fmoc-Phe-OH, 98%
Sigma-Aldrich
Fluorescein, for fluorescence, free acid
Sigma-Aldrich
L-Glutamine
Sigma-Aldrich
Dichloromethane, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Dichloromethane, suitable for HPLC, ≥99.9%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Stearic acid, reagent grade, 95%
Sigma-Aldrich
Stearic acid, Grade I, ≥98.5% (capillary GC)