Merck
  • Antimicrobial mechanism of resveratrol-trans-dihydrodimer produced from peroxidase-catalyzed oxidation of resveratrol.
Antimicrobial mechanism of resveratrol-trans-dihydrodimer produced from peroxidase-catalyzed oxidation of resveratrol.
Biotechnology and bioengineering (2015-06-26)
Mauricio Mora-Pale, Namita Bhan, Sayaka Masuko, Paul James, Julia Wood, Scott McCallum, Robert J Linhardt, Jonathan S Dordick, Mattheos A G Koffas
ABSTRACT
Plant polyphenols are known to have varying antimicrobial potencies, including direct antibacterial activity, synergism with antibiotics and suppression of bacterial virulence. We performed the in vitro oligomerization of resveratrol catalyzed by soybean peroxidase, and the two isomers (resveratrol-trans-dihydrodimer and pallidol) produced were tested for antimicrobial activity. The resveratrol-trans-dihydrodimer displayed antimicrobial activity against the Gram-positive bacteria Bacillus cereus, Listeria monocytogenes, and Staphylococcus aureus (minimum inhibitory concentration (MIC) = 15.0, 125, and 62.0 μM, respectively) and against Gram-negative Escherichia coli (MIC = 123 μM, upon addition of the efflux pump inhibitor Phe-Arg-β-naphthylamide). In contrast, pallidol had no observable antimicrobial activity against all tested strains. Transcriptomic analysis implied downregulation of ABC transporters, genes involved in cell division and DNA binding proteins. Flow cytometric analysis of treated cells revealed a rapid collapse in membrane potential and a substantial decrease in total DNA content. The active dimer showed >90% inhibition of DNA gyrase activity, in vitro, by blocking the ATP binding site of the enzyme. We thus propose that the resveratrol-trans-dihydrodimer acts to: (1) disrupt membrane potential; and (2) inhibit DNA synthesis. In summary, we introduce the mechanisms of action and the initial evaluation of an active bactericide, and a platform for the development of polyphenolic antimicrobials.
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Dichloromethane, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Dichloromethane, suitable for HPLC, ≥99.9%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Resveratrol, ≥99% (HPLC)
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Dichloromethane, biotech. grade, 99.9%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Dichloromethane, ACS reagent, ≥99.5%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)
Sigma-Aldrich
Ultrapure Acetonitrile
Sigma-Aldrich
Dichloromethane, contains 40-150 ppm amylene as stabilizer, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, for molecular biology
Sigma-Aldrich
Dichloromethane, ACS reagent, ≥99.5%, contains 40-150 ppm amylene as stabilizer
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Water, BioPerformance Certified
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
N,N-Dimethylformamide, anhydrous, 99.8%
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Sigma-Aldrich
Water, for cell biology, sterile ultrafiltered
Sigma-Aldrich
Water, for molecular biology, sterile filtered
Sigma-Aldrich
Anthraquinone, 97%