Merck
  • Changing in lipid profile induced by the mutation of Foxn1 gene: A lipidomic analysis of Nude mice skin.
Changing in lipid profile induced by the mutation of Foxn1 gene: A lipidomic analysis of Nude mice skin.
Biochimie (2015-10-03)
Justine Lanzini, Delphine Dargère, Anne Regazzetti, Abdellah Tebani, Olivier Laprévote, Nicolas Auzeil
ABSTRACT
Nude mice carry a spontaneous mutation affecting the gene Foxn1 mainly expressed in the epidermis. This gene is involved in several skin functions, especially in the proliferation and the differentiation of keratinocytes which are key cells of epithelial barrier. The skin, a protective barrier for the body, is essentially composed of lipids. Taking into account these factors, we conducted a lipidomic study to search for any changes in lipid composition of skin possibly related to Foxn1 mutation. Lipids were extracted from skin biopsies of Nude and BALB/c mice to be analyzed by liquid chromatography coupled to a high resolution mass spectrometer (HRMS). Multivariate and univariate data analyses were carried out to compare lipid extracts. Identification was performed using HRMS data, retention time and mass spectrometry fragmentation study. These results indicate that mutation of Foxn1 leads to significant modifications in the lipidome in Nude mice skin. An increase in cholesterol sulfate, phospholipids, sphingolipids and fatty acids associated with a decrease in glycerolipids suggest that the lipidome in mice skin is regulated by the Foxn1 gene.
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Methanol, NMR reference standard
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
tert-Butyl methyl ether, ACS reagent, ≥99.0%
Sigma-Aldrich
Chloroform, biotech. grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
tert-Butyl methyl ether, reagent grade, ≥98%
Sigma-Aldrich
Chloroform, contains 100-200 ppm amylenes as stabilizer, ≥99.5%