Merck
  • c-Src-dependent transactivation of EGFR mediates CORM-2-induced HO-1 expression in human tracheal smooth muscle cells.
c-Src-dependent transactivation of EGFR mediates CORM-2-induced HO-1 expression in human tracheal smooth muscle cells.
Journal of cellular physiology (2015-04-30)
Chuen-Mao Yang, Chih-Chung Lin, I-Ta Lee, Chih-Kai Hsu, Yu-Chen Tai, Hsi-Lung Hsieh, Pei-Ling Chi, Li-Der Hsiao
ABSTRACT
Carbon monoxide (CO), a reaction product of the cytoprotective heme oxygenase (HO)-1, displays an anti-inflammatory effect in various cellular injuries, but the precise mechanisms of HO-1 expression remain unknown. We used the transition metal carbonyl compound carbon monoxide-releasing molecule-2 (CORM-2) that acts as carbon monoxide donor. The effects of CORM-2 on expression of HO-1 in human tracheal smooth muscle cells (HTSMCs) were determined by Western blot, real-time PCR, and promoter activity assay. In HTSMCs, CORM-2 activated Nrf2 through the activation of a c-Src/EGFR/PI3K/Akt-dependent pathway, resulting in HO-1 expression. We showed that CORM-2-induced HO-1 protein and mRNA levels were inhibited by the inhibitor of c-Src (PP1 or SU6656), EGFR (AG1478), PI3K (LY294002), Akt (SH-5), JNK1/2 (SP600125), or p38 MAPK (SB202190) and transfection with siRNA of c-Src, EGFR, Akt, p38, JNK2, or Nrf2 in HTSMCs. We also showed that CORM-2 stimulated c-Src, EGFR, Akt, p38 MAPK, and JNK1/2 phosphorylation. CORM-2 also enhanced Nrf2 translocation from the cytosol to the nucleus and antioxidant response element (ARE) promoter activity. Moreover, CORM-2 mediated p38 MAPK and JNK1/2 activation via a c-Src/EGFR/PI3K/Akt pathway, which further enhanced Nrf2 activation and translocation. Finally, we observed that CORM-2 induced in vivo binding of Nrf2 to the HO-1 promoter. CORM-2 activates the c-Src/EGFR/PI3K/Akt/JNK1/2 and p38 MAPK pathways, which in turn trigger Nrf2 activation and ultimately induces HO-1 expression in HTSMCs. Thus, the HO-1/CO system might be potential therapeutics in airway diseases.
Sigma-Aldrich
Tricarbonyldichlororuthenium(II) dimer
Sigma-Aldrich
PP1, ≥98% (HPLC)
Sigma-Aldrich
SP600125, ≥98% (HPLC)
Sigma-Aldrich
MISSION® esiRNA, targeting human EGFR
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Egfr