Merck
  • In vitro antifungal activity and in vivo antibiofilm activity of cerium nitrate against Candida species.
In vitro antifungal activity and in vivo antibiofilm activity of cerium nitrate against Candida species.
The Journal of antimicrobial chemotherapy (2015-01-06)
Ana Silva-Dias, Isabel M Miranda, Joana Branco, Luís Cobrado, Matilde Monteiro-Soares, Cidália Pina-Vaz, Acácio G Rodrigues
ABSTRACT
The objective of this study was to clarify the antifungal properties of cerium, a lanthanide member, against Candida species. A comprehensive study with planktonic and sessile cells was performed. The ability of cerium nitrate (CN) to impair in vitro and in vivo biofilm formation was evaluated and its potential use in biofilm treatment was also evaluated. Forty-eight clinical isolates of different Candida species and the type strain ATCC 90028 were tested according to the protocol M27-A3. The MICs and minimum lethal concentrations were determined. A time-kill assay was performed and a cytometric kinetic study was performed using live/dead markers. Biofilm inhibition and biofilm susceptibility in the presence of cerium was evaluated by quantification of the biofilm metabolic activity and total biomass with XTT and crystal violet assays, respectively. CN in vivo efficacy as a coating for medical indwelling devices was evaluated for the first time for Candida parapsilosis, using a mouse subcutaneous foreign body model using polyurethane catheter segments. Scanning electron microscopy was used to assess biofilm architecture after CN treatment. The MICs for planktonic cells correlated with severe cellular metabolic activity impairment and membrane damage after 3 h of incubation. Moreover, CN efficiently prevented biofilm formation both in vitro and in vivo in segments of polyurethane catheters. At higher concentrations, it was also able to disorganize and almost eradicate preformed biofilms. Our results strongly suggest that CN application in the clinical setting might be effective in preventing the formation of biofilm-associated infections, namely through catheter coating and ultimately as an antimicrobial lock therapy.
Sigma-Aldrich
Sodium azide, purum p.a., ≥99.0% (T)
Sigma-Aldrich
MOPS, BioXtra, ≥99.5% (titration)
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Sodium azide, BioXtra
Sigma-Aldrich
MOPS, ≥99.5% (titration)
Sigma-Aldrich
MOPS, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Sigma-Aldrich
Sodium azide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
MOPS, BioPerformance Certified, suitable for cell culture, ≥99.5% (titration)
Sigma-Aldrich
Sodium azide, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Cerium(III) nitrate hexahydrate, 99.99% trace metals basis
Sigma-Aldrich
Cerium(III) nitrate hexahydrate, 99.999% trace metals basis
Sigma-Aldrich
Cerium, ingot, under oil, 99.9% trace rare earth metals basis
Sigma-Aldrich
Cerium, chips
Sigma-Aldrich
Cerium(III) nitrate hexahydrate, 99% trace metals basis
Sigma-Aldrich
Propidium iodide solution
Cerium, wire reel, 100mm, diameter 1.0mm, hard, 99.9%
Cerium, wire reel, 50mm, diameter 1.0mm, hard, 99.9%
Cerium, foil, not light tested, 25x25mm, thickness 0.025mm, as rolled, 99.9%
Cerium, foil, not light tested, 50x50mm, thickness 0.025mm, as rolled, 99.9%
Cerium, rod, 50mm, diameter 6.35mm, cast, 99.9%
Cerium, foil, 50x50mm, thickness 0.1mm, as rolled, 99.9%
Cerium, rod, 100mm, diameter 12.7mm, cast, 99.9%
Cerium, foil, not light tested, 25x25mm, thickness 0.005mm, as rolled, 99.9%
Cerium, rod, 100mm, diameter 6.35mm, cast, 99.9%
Cerium, rod, 50mm, diameter 12.7mm, cast, 99.9%
Cerium, foil, 25x25mm, thickness 0.1mm, as rolled, 99.9%
Cerium, foil, light tested, 25x25mm, thickness 0.25mm, as rolled, 99.9%
Cerium, wire reel, 25mm, diameter 1.0mm, hard, 99.9%
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)