Merck
  • Effects of liquid konjac on parameters related to obesity in diet-induced obese mice.
Effects of liquid konjac on parameters related to obesity in diet-induced obese mice.
Bioscience, biotechnology, and biochemistry (2015-04-03)
Seiichiro Aoe, Haruka Kudo, Seiya Sakurai
ABSTRACT
Konjac-mannan has been suggested to exert a protective effect against visceral obesity. However, there is little knowledge about the effects of liquid konjac (LK) that is partially alkali gelled. The purpose of the present work was to evaluate the potential beneficial effects of a LK powder on obesity in mice. Male C57BL/6J mice were fed a high-fat diet supplemented with either 2.5 or 5% LK powder for 80 days. Growth parameters, abdominal fat content, serum biochemical markers, and hepatic lipid accumulations were measured. Dietary supplementation with LK resulted in decreased body weight gain and abdominal fat accumulation. Dose-dependent decreases were observed in accumulation of hepatic lipids and serum total cholesterol, leptin, insulin concentrations. The study findings indicated that LK had preventing effect against obesity, including the reduction of abdominal and hepatic lipid accumulation and serum parameters related to obesity.
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Sigma-Aldrich
Cholesterol, from lanolin, ≥99.0% (GC)
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
SAFC
Cholesterol, from sheep wool, Controlled origin, meets USP/NF testing specifications
Sigma-Aldrich
Osmium tetroxide, Sealed ampule.
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, aqueous solution, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
Os EnCat® 40, extent of labeling: 0.3 mmol/g Os loading
Sigma-Aldrich
Glycerol, BioUltra, for molecular biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Glycerol, FCC, FG
SAFC
Cholesterol, Plant-Derived, SyntheChol®
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Osmium tetroxide solution, suitable for electron microscopy, 2% in H2O
Sigma-Aldrich
2-Propanol, for molecular biology, BioReagent, ≥99.5%
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Osmium tetroxide solution, suitable for electron microscopy, 4% in H2O
Sigma-Aldrich
Glycerol, for molecular biology, ≥99.0%
Sigma-Aldrich
2-Propanol, anhydrous, 99.5%
Sigma-Aldrich
2-Propanol, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%, poly coated bottles
Sigma-Aldrich
2-Propanol, electronic grade, 99.999% trace metals basis