Merck
  • Kif20a inhibition reduces migration and invasion of pancreatic cancer cells.
Kif20a inhibition reduces migration and invasion of pancreatic cancer cells.
The Journal of surgical research (2015-05-09)
Daniela Stangel, Mert Erkan, Malte Buchholz, Thomas Gress, Christoph Michalski, Susanne Raulefs, Helmut Friess, Jörg Kleeff
ABSTRACT
The Translational Genome Research Network in Pancreatic Cancer performed a meta-analysis of publicly available various high-throughput gene analysis panels to identify drugable targets. There, the most differentially expressed gene between normal and cancerous pancreas was Kif20a. The aim of the study was to verify this expression pattern and further characterize Kif20a in pancreatic cancer. Detailed expression analyses were carried out in pancreatic tissues and in a wide panel of pancreatic cells including ductal adenocarcinoma (PDAC) and neuroendocrine-cancer cell lines as well as immortalized human pancreatic ductal epithelial and primary stellate cells using quantitative real-time polymerase chain reaction, immunohistochemistry, immunofluorescence, and immunoblot analyses. Effects on proliferation, apoptosis, and cell cycle were assessed by MTT assays, caspase-cleavage assays, and fluorescence-activated cell sorting analysis after Kif20a silencing. Cell motility was assessed by migration and invasion assays as well as time-lapse microscopy. Mean Kif20a messenger RNA expression was 18.4-fold upregulated in PDAC tissues compared with that in the normal pancreas. In line, neuroendocrine-cancer cell lines display a 1.6-fold increase and ductal adenocarcinoma cell lines a 11-fold increase of Kif20a messenger RNA (P = 0.009) in comparison with primary stellate cells. A 7.3-fold overexpression was also found in immortalized pancreatic ductal epithelial cells. Kif20a silencing with small interfering RNA molecules resulted in an inhibition of proliferation, motility, and invasion of pancreatic cancer cell lines. Targeting Kif20a reduces proliferation, migration, and invasion of pancreatic cancer cells. Together with its significant overexpression in PDAC, this makes it a potential target for diagnostic and interventional purposes.
Sigma-Aldrich
Thymidine, ≥99.0% (HPLC)
Sigma-Aldrich
Guanine, 98%
Sigma-Aldrich
Thymidine, ≥99%
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Thymidine, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Guanine, BioUltra
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Kif20a
Sigma-Aldrich
MISSION® esiRNA, targeting human KIF20A
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%