Merck
  • Design, Preparation, and Characterization of Zn and Cu Metallopeptides Based On Tetradentate Aminopyridine Ligands Showing Enhanced DNA Cleavage Activity.
Design, Preparation, and Characterization of Zn and Cu Metallopeptides Based On Tetradentate Aminopyridine Ligands Showing Enhanced DNA Cleavage Activity.
Inorganic chemistry (2015-10-28)
Marta Soler, Eduard Figueras, Joan Serrano-Plana, Marta González-Bártulos, Anna Massaguer, Anna Company, Ma Ángeles Martínez, Jaroslav Malina, Viktor Brabec, Lidia Feliu, Marta Planas, Xavi Ribas, Miquel Costas
ABSTRACT
The conjugation of redox-active complexes that can function as chemical nucleases to cationic tetrapeptides is pursued in this work in order to explore the expected synergistic effect between these two elements in DNA oxidative cleavage. Coordination complexes of biologically relevant first row metal ions, such as Zn(II) or Cu(II), containing the tetradentate ligands 1,4-dimethyl-7-(2-pyridylmethyl)-1,4,7-triazacyclononane ((Me2)PyTACN) and (2S,2S')-1,1'-bis(pyrid-2-ylmethyl)-2,2'-bipyrrolidine ((S,S')-BPBP) have been linked to a cationic LKKL tetrapeptide sequence. Solid-phase synthesis of the peptide-tetradentate ligand conjugates has been developed, and the preparation and characterization of the corresponding metallotetrapeptides is described. The DNA cleavage activity of Cu and Zn metallopeptides has been evaluated and compared to their metal binding conjugates as well as to the parent complexes and ligands. Very interestingly, the oxidative Cu metallopeptides 1Cu and 2Cu show an enhanced activity compared to the parent complexes, [Cu(PyTACN)](2+) and [Cu(BPBP)](2+), respectively. Under optimized conditions, 1Cu displays an apparent pseudo first-order rate constant (kobs) of ∼0.16 min(-1) with a supercoiled DNA half-life time (t1/2) of ∼4.3 min. On the other hand, kobs for 2Cu has been found to be ∼0.11 min(-1) with t1/2 ≈ 6.4 min. Hence, these results point out that the DNA cleavage activities promoted by the metallopeptides 1Cu and 2Cu render ∼4-fold and ∼23 rate accelerations in comparison with their parent Cu complexes. Additional binding assays and mechanistic studies demonstrate that the enhanced cleavage activities are explained by the presence of the cationic LKKL tetrapeptide sequence, which induces an improved binding affinity to the DNA, thus bringing the metal ion, which is responsible for cleavage, in close proximity.
Sigma-Aldrich
Tetrahydrofuran, anhydrous, ≥99.9%, inhibitor-free
Sigma-Aldrich
Diethyl ether, contains 1 ppm BHT as inhibitor, anhydrous, ≥99.7%
Sigma-Aldrich
Methyl Green, zinc chloride salt, certified by the BSC, Dye content 85 %
Sigma-Aldrich
tert-Butylchlorodimethylsilane solution, 50 wt. % in toluene
Sigma-Aldrich
N,N-Dimethylformamide, anhydrous, 99.8%
Sigma-Aldrich
Piperidine, ReagentPlus®, 99%
Sigma-Aldrich
Tetrabutylammonium fluoride on silica gel, ~1.5 mmol/g (F-)
Sigma-Aldrich
Diethyl ether
Supelco
Tetrahydrofuran, HPLC grade, ≥99.9%, inhibitor-free
Sigma-Aldrich
Sodium borohydride solution, 2.0 M in triethylene glycol dimethyl ether
Sigma-Aldrich
Methyl Green, zinc chloride salt, <0.5% crystal violet, Dye content 80 %
Sigma-Aldrich
Ammonia solution, 0.4 M in THF
Sigma-Aldrich
Sodium borohydride solution, ~12 wt. % in 14 M NaOH
Sigma-Aldrich
Ammonia, anhydrous, ≥99.98%
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
(+)-Sodium L-ascorbate, crystalline, ≥98%
Sigma-Aldrich
(+)-Sodium L-ascorbate, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Methyl Green, zinc chloride salt, ~85%
Sigma-Aldrich
Dimethyl 2,5-pyridine dicarboxylate, 97%
Supelco
Pyridaben, PESTANAL®, analytical standard
Sigma-Aldrich
Sodium azide, purum p.a., ≥99.0% (T)
Sigma-Aldrich
(+)-Sodium L-ascorbate, BioXtra, ≥99.0% (NT)
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Tetrahydrofuran, anhydrous, contains 250 ppm BHT as inhibitor, ≥99.9%
Sigma-Aldrich
Ammonia-14N, 99.99 atom % 14N
Sigma-Aldrich
VenPure® SF, powder
Sigma-Aldrich
Sodium azide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Diethyl ether, ACS reagent, ≥98.0%, contains ≤2% ethanol and ≤10ppm BHT as inhibitor