Merck
  • miRNA-24-3p promotes cell proliferation and inhibits apoptosis in human breast cancer by targeting p27Kip1.
miRNA-24-3p promotes cell proliferation and inhibits apoptosis in human breast cancer by targeting p27Kip1.
Oncology reports (2015-06-06)
Kangping Lu, Jingxuan Wang, Ying Song, Shu Zhao, Hang Liu, Dabei Tang, Bo Pan, Hong Zhao, Qingyuan Zhang
ABSTRACT
MicroRNAs (miRNAs) are often aberrantly expressed in breast cancer and are postulated to play a role in its initiation and progression. In the present study, we found that the expression level of miR-24-3p was upregulated in breast cancer in comparison with the level in adjacent normal tissues. Overexpression of miR-24-3p was able to promote cell proliferation and inhibit cell apoptosis in MDA-MB-435 and MDA-MB-468 cells. With the bioinformatic method, we further identified that p27Kip1 is a direct target of miR-24-3p, and its protein level was negatively regulated by miR-24-3p. Therefore, the data reported here demonstrate that miR-24-3p is an important regulator in breast cancer, and imply that the miR-24-3p/p27Kip1 axis has potential as a therapeutic target for breast cancer.
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Psma6
Sigma-Aldrich
MISSION® esiRNA, targeting human PSMA6, RP11-561B11.2
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%