Merck
MiR-212-3p inhibits glioblastoma cell proliferation by targeting SGK3.
Journal of neuro-oncology (2015-02-28)
Huailei Liu, Chenguang Li, Chen Shen, Fei Yin, Kaikai Wang, Yaohua Liu, Bingjie Zheng, Weiguang Zhang, Xu Hou, Xin Chen, Jianing Wu, Xiaoxiong Wang, Chen Zhong, Jiakang Zhang, Huaizhang Shi, Jing Ai, Shiguang Zhao
ABSTRACT
Glioblastoma multiforme (GBM) is the most malignant brain tumor in humans. Previous studies have demonstrated that microRNA plays important roles in the development and proliferation of GBM cells. Here we defined the mechanism by which miR-212-3p regulated the proliferation of GBM. In this study, we showed that miR-212-3p expression was significantly down-regulated and negatively correlated with serum and glucocorticoid-inducible kinase 3 (SGK3) in GBM. Either over-expression of miR-212-3p or silence of SGK3 decreased viability of GBM cells. Moreover, miR-212-3p directly bound to 3'UTR of SGK3 and inhibited its mRNA and protein expression. And over-expression of SGK3 rescued the decreased proliferation of GBM cells induced by miR-212-3p. Importantly, miR-212-3p also suppressed tumor growth in vivo. Collectively, our results demonstrated that miR-212-3p inhibited proliferation of GBM cells by directly targeting SGK3, and could potentially serve as a new therapeutic target for GBM.
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
MISSION® esiRNA, targeting human SGK3, C8ORF44-SGK3
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Sgk3
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®