Merck
In Situ Detection of Particle Aggregation on Electrode Surfaces.
Chemphyschem : a European journal of chemical physics and physical chemistry (2015-06-04)
Kamonwad Ngamchuea, Kristina Tschulik, Shaltiel Eloul, Richard G Compton
ABSTRACT
Partially blocked electrodes (PBEs) are important; many applications use non-conductive nanoparticles (NPs) to introduce new electrode functionalities. As aggregation is a problem in NP immobilization, developing an in situ method to detect aggregation is vital to characterise such modified electrodes. We present chronoamperometry as a method for detection of NP surface aggregation and semi-quantitative sizing of the formed aggregates, based on the diffusion limited current measured at PBEs as compared with the values calculated numerically for different blocking feature sizes. In contrast to voltammetry, no approximations on electrode kinetics are needed, making chronoamperometry a more general and reliable method. Sizing is shown for two modification methods. Upon drop casting, significant aggregation is observed, while it is minimized in electrophoretic NP deposition. The aggregate sizes determined are in semi-quantitative agreement with ex situ microscopic analysis of the PBEs.
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Potassium nitrate, BioXtra, ≥99.0%
Sigma-Aldrich
Potassium nitrate, BioReagent, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Potassium nitrate, 99.99% trace metals basis
Sigma-Aldrich
Potassium nitrate, 99.999% trace metals basis
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
Potassium hexacyanoferrate(II) trihydrate, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
Potassium hexacyanoferrate(III), BioUltra, ≥99.0% (RT)
Sigma-Aldrich
Hydrogen chloride, ReagentPlus®, ≥99%
Sigma-Aldrich
Potassium hexacyanoferrate(III), 99.98% trace metals basis
Sigma-Aldrich
Potassium hexacyanoferrate(II) trihydrate, ≥99.95% trace metals basis