Merck
  • ERK5 regulates basic fibroblast growth factor-induced type 1 plasminogen activator inhibitor expression and cell proliferation in lung fibroblasts.
ERK5 regulates basic fibroblast growth factor-induced type 1 plasminogen activator inhibitor expression and cell proliferation in lung fibroblasts.
Life sciences (2015-06-03)
Jung-Hwa Han, Ae-Rang Hwang, Dae-Hwan Nam, Suji Kim, Hyoung Chul Choi, Jae Hyang Lim, Chang-Hoon Woo
ABSTRACT
bFGF is a potent mitogen of cells associated with fibrosis. Although ERK5 has been reported to play roles in the development of fibrosis, its roles in regulating bFGF-induced fibrotic responses are not understood, especially in lung fibroblasts. The authors investigated the role of ERK5 in bFGF induction of cell proliferation and in induction of PAI-1, a critical regulator of the pathological features of fibrosis, in lung fibroblasts. The role played by ERK5 in bFGF-induced PAI-1 expression was elucidated by perturbing the ERK5 signaling pathway using a specific chemical inhibitor and siRNA of ERK5. The effects of ERK5 signal perturbation on PAI-1 expression were measured at multiple levels by Q-PCR, immunoblotting, ELISA, and reporter gene analysis. The role of MEF2 in bFGF-induced activation of PAI-1 promoter activity via ERK5 was measured using a biotin-labeled DNA pull-down assay, and the effects of ERK5 on the mitogenic effects of bFGF were assessed using a MTT assay. In both primary human lung fibroblast and lung fibroblast cell lines, inhibition of ERK5 blocked bFGF-induced PAI-1 expression at both mRNA and protein levels and inhibited bFGF-induced PAI-1 promoter activity induction by bFGF. Upon stimulation with bFGF, MEF2 directly bound to the consensus sequence of the MEF2 binding site in the PAI-1 promoter. In addition, bFGF-induced PAI-1 up-regulation was inhibited by MEF2 siRNA, and bFGF-induced fibroblast proliferation was blocked by inhibiting ERK5. This study reveals a novel role for the ERK5-MEF2 cascade, linking bFGF-induced PAI-1 expression and subsequent mitogenic processes in lung fibroblasts.
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
MISSION® esiRNA, targeting human MEF2D
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
FGF-2 human, recombinant, expressed in E. coli, ≥95% (SDS-PAGE), ≥95% (HPLC)
Sigma-Aldrich
MISSION® esiRNA, targeting human MEF2C
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Mef2d
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
FGF-2 human, recombinant, expressed in insect cells, ≥85% (SDS-PAGE)
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O