Merck
  • MnCo2O4 nanowires anchored on reduced graphene oxide sheets as effective bifunctional catalysts for Li-O2 battery cathodes.
MnCo2O4 nanowires anchored on reduced graphene oxide sheets as effective bifunctional catalysts for Li-O2 battery cathodes.
ChemSusChem (2015-04-25)
Jong Guk Kim, Youngmin Kim, Yuseong Noh, Won Bae Kim
ABSTRACT
A hybrid composite system of MnCo2 O4 nanowires (MCO NWs) anchored on reduced graphene oxide (RGO) nanosheets was prepared as the bifunctional catalyst of a Li-O2 battery cathode. The catalysts can be obtained from the hybridization of one-dimensional MCO NWs and two-dimensional RGO nanosheets. As O2 -cathode catalysts for Li-O2 cells, the MCO@RGO composites showed a high initial discharge capacity (ca. 11092.1 mAh gcarbon (-1) ) with a high rate performance. The Li-O2 cells could run for more than 35 cycles with high reversibility under a limited specific capacity of 1000 mAh gcarbon (-1) with a low potential polarization of 1.36 V, as compared with those of pure Ketjenblack and MCO NWs. The high cycling stability, low potential polarization, and rate capability suggest that the MCO@RGO composites prepared here are promising catalyst candidates for highly reversible Li-O2 battery cathodes.
Sigma-Aldrich
Oxalic acid, ReagentPlus®, ≥99%
Sigma-Aldrich
Oxalic acid, 98%
Sigma-Aldrich
Lithium, wire (in mineral oil), diam. 3.2 mm, 99.9% trace metals basis
Sigma-Aldrich
Lithium, ribbon, thickness × W 0.75 mm × 19 mm, 99.9% trace metals basis
Sigma-Aldrich
Lithium, wire, diam. 3.2 mm, in mineral oil, ≥98%
Sigma-Aldrich
Oxalic acid, puriss. p.a., anhydrous, ≥99.0% (RT)
Sigma-Aldrich
Lithium, ribbon, thickness × W 1.5 mm × 100 mm, 99.9% trace metals basis
Sigma-Aldrich
Lithium, rod, diam. 12.7 mm, 99.9% trace metals basis
Sigma-Aldrich
Lithium, granular, 99% trace metals basis
Sigma-Aldrich
Lithium, ribbon, thickness × W 0.75 mm × 45 mm, 99.9% trace metals basis
Sigma-Aldrich
Lithium, ribbon, thickness × W 0.38 mm × 23 mm, 99.9% trace metals basis
Sigma-Aldrich
Lithium, granular, 4-10 mesh particle size, high sodium, 99% (metals basis)
Sigma-Aldrich
Oxalic acid, purified grade, 99.999% trace metals basis
Lithium, foil, 25x100mm, thickness 0.6mm, as rolled, 99.9%
Lithium, foil, not light tested, 38x200mm, thickness 0.20mm, as rolled, 99.9%
Sigma-Aldrich
Lithium, foil, not light tested, 38x500mm, thickness 0.20mm, as rolled, 99.9%
Sigma-Aldrich
Cyclohexane, anhydrous, 99.5%
Sigma-Aldrich
1-Pentanol, puriss. p.a., ACS reagent, ≥99.0% (GC)
Sigma-Aldrich
Amyl alcohol, ≥99%, FG
Sigma-Aldrich
1-Pentanol, ACS reagent, ≥99%
Sigma-Aldrich
Lithium-7Li, ≥99.8 atom % 7Li, ≥99.8% (CP)
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, BioXtra, ≥99%
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, ≥98%
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, BioUltra, for molecular biology, ≥99.0% (AT)
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, for molecular biology, ≥99%
SAFC
Hexadecyltrimethylammonium bromide, USP/NF