Merck
  • The molecular mechanism underlying the induction of hepatic MRP3 expression and function by omeprazole.
The molecular mechanism underlying the induction of hepatic MRP3 expression and function by omeprazole.
Biopharmaceutics & drug disposition (2015-01-20)
Yu-Qin Pan, Qiong-Yu Mi, Bang-Shun He, Shu-Li Zhao, Ting Tai, Hong-Guang Xie
ABSTRACT
Previous work has indicated that there is increased protein expression of multidrug resistance-associated protein 3 (MRP3) in the liver samples of patients treated with omeprazole compared with those who were not. However, evidence is still lacking to show the mechanisms underlying that induction. This study aimed to assess changes in the fold-induction of MRP3 mRNA and protein expression over controls in omeprazole-treated HepG2 cells after transient transfection of human MRP3 siRNA, or after pretreatment with actinomycin D (Act-D). Furthermore, MRP3 siRNA knock-down or MRP-specific inhibition (indomethacin) was used to determine whether the MRP3 protein induced by omeprazole possessed an enhanced efflux transport. The results demonstrated that omeprazole induced MRP3 mRNA and protein expression in a concentration- and time-dependent manner. Moreover, that induction was almost completely abolished by the addition of human MRP3 siRNA and also by pretreatment with Act-D, respectively. In addition, the decay rate of MRP3 mRNA in vehicle- and omeprazole-treated cells was similar in the presence of Act-D, suggesting transcriptional up-regulation of MRP3 mRNA expression by omeprazole. Most importantly, omeprazole induced MRP3 efflux transport activity, as measured by the 5-carboxyfluorescein assay in the absence and presence of human MRP3 siRNA or indomethacin. It is concluded that omeprazole can induce MRP3 mRNA and protein expression and enhance MRP3 efflux transport activity through transcriptional up-regulation, and that omeprazole can also induce other MRP transporters.
Sigma-Aldrich
MISSION® esiRNA, targeting human ABCC3
Sigma-Aldrich
Actinomycin D, from Streptomyces sp., ~98% (HPLC)
Sigma-Aldrich
(Tyr[SO3H]27)Cholecystokinin fragment 26-33 Amide, ≥97% (HPLC), powder
Sigma-Aldrich
Acrylamide, suitable for electrophoresis, ≥99% (HPLC), powder
Sigma-Aldrich
Acrylamide, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Actinomycin D, from Streptomyces sp., ≥95% (HPLC)
Sigma-Aldrich
Actinomycin D, from Streptomyces sp., suitable for cell culture, ≥95%
Sigma-Aldrich
Indomethacin, meets USP testing specifications
Sigma-Aldrich
Indomethacin, 98.5-100.5% (in accordance with EP)
Sigma-Aldrich
Acrylamide, ≥99% (HPLC)
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Omeprazole, solid
Sigma-Aldrich
Dimethyl sulfoxide
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Acrylamide solution, 40%, suitable for electrophoresis, sterile-filtered
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Abcc3