Merck
  • Relationships between Chemical Characteristics and Phytotoxicity of Biochar from Poultry Litter Pyrolysis.
Relationships between Chemical Characteristics and Phytotoxicity of Biochar from Poultry Litter Pyrolysis.
Journal of agricultural and food chemistry (2015-07-08)
Alessandro G Rombolà, Giovanni Marisi, Cristian Torri, Daniele Fabbri, Alessandro Buscaroli, Michele Ghidotti, Andreas Hornung
ABSTRACT
Three biochars were prepared by intermediate pyrolysis from poultry litter at different temperatures (400, 500, and 600 °C with decreasing residence times) and compared with biochars from corn stalk prepared under the same pyrolysis conditions. The phytotoxicity of these biochars was estimated by means of seed germination tests on cress (Lepidium sativum L.) conducted in water suspensions (at 2, 5, and 40 g/L) and on biochars wetted according to their water-holding capacity. Whereas the seeds germinated after 72 h in water suspensions with corn stalk biochar were similar to the control (water only), significant inhibition was observed with poultry litter biochars. In comparison to corn stalk, poultry litter generated biochars with higher contents of ash, ammonium, nitrogen, and volatile fatty acids (VFAs) and a similar concentration of polycyclic aromatic hydrocarbons (PAHs). Results from analytical pyrolysis (Py-GC-MS) indicated that nitrogen-containing organic compounds (NCCs) and aliphatic components were distinctive constituents of the thermally labile fraction of poultry litter biochar. The inhibition of germination due to poultry litter biochar produced at 400 °C (PL400) was suppressed after solvent extraction or treatment with active sludge. A novel method based on solid-phase microextraction (SPME) enabled the identification of mobile organic compounds in PL400 capable of being released in air and water, including VFAs and NCCs. The higher phytotoxicity of poultry litter than corn biochars was tentatively attributed to hydrophilic biodegradable substances derived from lipids or proteins removable by water leaching or microbial treatments.
Product Description
Supelco
SPME fiber assembly polyacrylate (PA), df 85 μm(PA, for use with manual holder, needle size 24 ga
Supelco
SPME Portable Field Sampler, coating PDMS/DVB
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 7 μm(PDMS, needle size 24 ga, for use with manual holder
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 7 μm(PDMS, needle size 24 ga, for use with autosampler
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 100 μm(PDMS, for use with autosampler, needle size 23 ga
Supelco
SPME fiber assembly polyacrylate (PA), df 85 μm(PA, for use with autosampler, needle size 24 ga
Supelco
SPME Fiber Assembly Polydimethylsiloxane/Divinylbenzene (PDMS/DVB), df 65 μm(PDMS/DVB, needle size 24 ga, StableFlex, for use with manual holder
Supelco
SPME Fiber Assembly Polydimethylsiloxane/Divinylbenzene (PDMS/DVB), fused silica fiber, df 65 μm(PDMS/DVB, for use with manual holder, needle size 23 ga
Supelco
SPME Sampling Stand, for use with 4 mL vials
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 100 μm(PDMS, for use with autosampler, needle size 24 ga
Supelco
SPME Fiber Assembly Polydimethylsiloxane/Divinylbenzene (PDMS/DVB), df 65 μm(PDMS/DVB, needle size 24 ga, StableFlex, for use with autosampler
Supelco
SPME Portable Field Sampler, coating CAR/PDMS
Supelco
SPME Sampling Stand, for use with 15 mL vials
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 30 μm(PDMS, needle size 24 ga, for use with manual holder
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 30 μm(PDMS, needle size 24 ga, for use with autosampler
Supelco
SPME Fiber Assembly Polydimethylsiloxane/Divinylbenzene (PDMS/DVB), df 65 μm(PDMS/DVB, needle size 24 ga, for use with manual holder
Supelco
SPME Fiber Assembly Polydimethylsiloxane/Divinylbenzene (PDMS/DVB), df 65 μm(PDMS/DVB, for use with autosampler, needle size 23 ga
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 100 μm(PDMS, for use with manual holder, needle size 23 ga
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 7 μm(PDMS, needle size 23 ga, for use with autosampler
Supelco
SPME fiber assembly polyacrylate (PA), df 85 μm(PA, for use with autosampler, needle size 23 ga
Supelco
SPME Fiber Assembly Polydimethylsiloxane/Divinylbenzene (PDMS/DVB), df 65 μm(PDMS/DVB, needle size 23 ga, PDMS/DVB StableFlex, for use with autosampler
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 100 μm(PDMS, needle size 24 ga, for use with manual holder
Supelco
SPME Portable Field Sampler, coating PDMS
Supelco
SPME fiber assembly, Carbowax-Polyethylene Glycol (PEG) Coating, needle size 23 ga, df 60 μm(PEG, for use with autosampler
Supelco
SPME fiber assembly, Carbowax-Polyethylene Glycol (PEG) Coating, needle size 23 ga, df 60 μm(PEG, metal alloy fiber, for use with manual holder
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), metal alloy fiber, df 100 μm(PDMS, needle size 23 ga, for use with autosampler
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Nonane, anhydrous, ≥99%
Sigma-Aldrich
Acenaphthene-d10, 99 atom % D
Sigma-Aldrich
Cyclohexane, anhydrous, 99.5%