Merck
  • Dunaliella tertiolecta (Chlorophyta) Avoids Cell Death Under Ultraviolet Radiation By Triggering Alternative Photoprotective Mechanisms.
Dunaliella tertiolecta (Chlorophyta) Avoids Cell Death Under Ultraviolet Radiation By Triggering Alternative Photoprotective Mechanisms.
Photochemistry and photobiology (2015-08-01)
María Segovia, Teresa Mata, Armando Palma, Candela García-Gómez, Rosario Lorenzo, Alicia Rivera, Félix L Figueroa
ABSTRACT
The effect of different ultraviolet radiation (UVR) treatments combining PAR (P), UVA (A) and UVB (B) on the molecular physiology of Dunaliella tertiolecta was studied during 6 days to assess the response to chronic UVR exposure. UVR reduced cell growth but did not cause cell death, as shown by the absence of SYTOX Green labeling and cellular morphology. However, caspase-like enzymatic activities (CLs), (regarded as cell death proteases), were active even though the cells were not dying. Maximal quantum yield of fluorescence (Fv /Fm ) and photosynthetic electron transport rate (ETR) dropped. Decreased nonphotochemical quenching (NPQ) paralleled a drop in xanthophyll cycle de-epoxidation under UVB. Reactive oxygen species (ROS) and D1 protein accumulation were inversely correlated. PAB exhibited elevated ROS production at earlier times. Once ROS decayed, D1 protein recovered two-fold compared with P and PA at later stages. Therefore, PsbA gene was still transcribed, suggesting ROS involvement in D1 recovery by its direct effect on mRNA-translation. We add evidence of an UVB-induced positive effect on the cells when P is present, providing photoprotection and resilience, by means of D1 repair. This allowed cells to survive. The photoprotective mechanisms described here (which are counterintuitive in principle) conform to an important ecophysiological response regarding light stress acclimation.
Product Description
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, 2 M in H2O
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~0.025 M in H2O
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
3-[(3-Cholamidopropyl)dimethylammonio]-1-propanesulfonate hydrate, 98%
Sigma-Aldrich
Chloroform, ReagentPlus®, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, contains 100-200 ppm amylenes as stabilizer, ≥99.5%
Sigma-Aldrich
Chloroform, biotech. grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, ACS spectrophotometric grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, meets analytical specification of BP, 99-99.4% (GC)
Sigma-Aldrich
Chloroform, contains amylenes as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Magnesium chloride solution, for molecular biology, 1.00 M±0.01 M
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
Magnesium chloride solution, 0.1 M
Sigma-Aldrich
Magnesium chloride, anhydrous, ≥98%
Sigma-Aldrich
Magnesium chloride, BioReagent, suitable for insect cell culture, ≥97.0%
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
2-Propanol, puriss. p.a., ACS reagent, ≥99.8% (GC)
Sigma-Aldrich
Isopropyl alcohol, meets USP testing specifications
Sigma-Aldrich
2-Propanol, Laboratory Reagent, ≥99.5%
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Sigma-Aldrich
2-Propanol, for molecular biology, BioReagent, ≥99.5%