Merck
  • A modified m-CP medium for enumerating Clostridium perfringens from water samples.
A modified m-CP medium for enumerating Clostridium perfringens from water samples.
Canadian journal of microbiology (1988-01-01)
R Armon, P Payment
ABSTRACT
Medium m-CP, designed for the isolation of Clostridium perfringens from water samples, contains indoxyl beta-D-glucoside, an expensive chemical that is present at a high concentration in this medium. The use of m-CP with three concentrations of indoxyl beta-D-glucoside was tested at 0, 60, and 600 mg/L. Lowering the amount of indoxyl beta-D-glucoside to 60 mg/L (1/10 the recommended concentration) reduced the cost of this medium without affecting its sensitivity.
Product Description
Millipore
m-CP Agar Base, suitable for microbiology, NutriSelect® Plus
Sigma-Aldrich
Indoxyl β-D-glucoside, ≥97%