Merck
  • Urine proteome analysis to evaluate protein biomarkers in children with autism.
Urine proteome analysis to evaluate protein biomarkers in children with autism.
Clinica chimica acta; international journal of clinical chemistry (2015-08-25)
Vijayashankar Suganya, Arumugam Geetha, Sridharan Sujatha
ABSTRACT
Autism is a complex developmental disability for which no specific diagnostic markers have been identified so far. The present study aimed to evaluate whether there is any abnormal protein(s) excreted in the urine of autistic children by proteome analysis which may act as diagnostic marker. Urine proteome analysis was carried out in first void urine samples of autistic and normal children (n=30) in the age group of 4-12 years by 2D-PAGE followed by MALDI-TOF-MS analysis. Comparison of 2D-PAGE gels revealed that many urinary proteins are expressed differentially in autistic children. Total numbers of spots observed were 250 and 159 in autism and normal samples respectively, out of which 95 matches were observed. In addition, 3 spots of abnormally expressed peptides were selected, excised and analyzed. Peptide sequence with significant match score was for kininogen-1 (KNG-1)-50 (spot-1), IgG1 heavy chain variable region-35(spot-2) and mannan-binding lectin serine protease-2 isoform-2 precursor-45(spot-3). All the autistic children showed significant increase (p<0.001) in urinary kininogen level measured quantitatively by ELISA, when compared to normal children. Increased urinary kininogen-1 level in all the autistic children and the possibility of this protein as a diagnostic marker need further investigation.
Sigma-Aldrich
Glycine, ACS reagent, ≥98.5%
Sigma-Aldrich
Urea-12C, 99.9 atom % 12C
Sigma-Aldrich
Glycine, BioUltra, for molecular biology, ≥99.0% (NT)
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Acetone, ≥99%, meets FCC analytical specifications
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, for molecular biology, ~1 M in H2O
SAFC
Glycine
Sigma-Aldrich
3-[(3-Cholamidopropyl)dimethylammonio]-1-propanesulfonate hydrate, 98%
Sigma-Aldrich
Glycine, 99%, FCC
Sigma-Aldrich
Glycine, puriss. p.a., reag. Ph. Eur., buffer substance, 99.7-101% (calc. to the dried substance)
Sigma-Aldrich
Glycine, meets analytical specification of Ph. Eur., BP, USP, 99-101% (based on anhydrous substance)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
N,N′-Methylenebis(acrylamide), 99%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Thiourea, ACS reagent, ≥99.0%
Sigma-Aldrich
Acrylamide, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Urea solution, 40 % (w/v) in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Thiourea, ReagentPlus®, ≥99.0%
Sigma-Aldrich
N,N′-Methylenebisacrylamide, suitable for electrophoresis (after filtration or allowing insolubles to settle)
Sigma-Aldrich
Glycine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
Sigma-Aldrich
Glycine, BioXtra, ≥99% (titration)
Sigma-Aldrich
N,N′-Methylenebisacrylamide, powder, for molecular biology, suitable for electrophoresis, ≥99.5%
Sigma-Aldrich
Glycine, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Glycine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Acrylamide, suitable for electrophoresis, ≥99% (HPLC), powder