Merck
  • Influence of pesticide use in fruit orchards during blooming on honeybee mortality in 4 experimental apiaries.
Influence of pesticide use in fruit orchards during blooming on honeybee mortality in 4 experimental apiaries.
The Science of the total environment (2015-09-24)
Pau Calatayud-Vernich, Fernando Calatayud, Enrique Simó, Maria Morales Suarez-Varela, Yolanda Picó
ABSTRACT
Samples of dead honey bees (Apis mellifera L.) were collected periodically from 4 different locations during citrus and stone fruit trees blooming season to evaluate the potential impact of agrochemicals on honey bee death rate. For the determination of mortality, dead honey bee traps were placed in front of the experimental hives entrance located in areas of intensive agriculture in Valencian Community (Spain). A total of 34 bee samples, obtained along the monitoring period, were analyzed by means of QuEChERS extraction method and screened for 58 pesticides or their degradation products by LC-MS/MS. An average of four pesticides per honey bee sample was detected. Coumaphos, an organophosphate acaricide used against varroosis in the experimental hives, was detected in 94% of the samples. However, this acaricide was unlikely to be responsible for honey bee mortality because its constantly low concentration during all the monitoring period, even before and after acute mortality episodes. The organophosphates chlorpyrifos and dimethoate, as well as the neonicotinoid imidacloprid, were the most frequently detected agrochemicals. Almost 80% of the samples had chlorpyrifos, 68% dimethoate, and 32% imidacloprid. Maximum concentrations for these three compounds were 751, 403, 223 ng/g respectively. Influence of these pesticides on acute honey bee mortality was demonstrated by comparing coincidence between death rate and concentrations of chlorpyrifos, dimethoate and imidacloprid.
Sigma-Aldrich
Ammonium formate solution, BioUltra, 10 M in H2O
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O
Supelco
Atrazine, analytical standard
Sigma-Aldrich
Magnesium sulfate solution, BioUltra, for molecular biology
Sigma-Aldrich
Magnesium sulfate, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Magnesium sulfate solution, for molecular biology, 1.00 M±0.04 M
Sigma-Aldrich
5-(N,N-Dimethyl)amiloride hydrochloride
Sigma-Aldrich
Magnesium sulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Ultrapure Acetonitrile
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Magnesium sulfate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Magnesium sulfate, puriss. p.a., drying agent, anhydrous, ≥98.0% (KT), powder (very fine)
Sigma-Aldrich
Magnesium sulfate, anhydrous, free-flowing, Redi-Dri, reagent grade, ≥97%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Magnesium sulfate, anhydrous, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Thiabendazole, suitable for plant cell culture