Merck
  • Kinetic analysis of the reduction of 4-nitrophenol catalyzed by Au/Pd nanoalloys immobilized in spherical polyelectrolyte brushes.
Kinetic analysis of the reduction of 4-nitrophenol catalyzed by Au/Pd nanoalloys immobilized in spherical polyelectrolyte brushes.
Physical chemistry chemical physics : PCCP (2015-03-20)
Sasa Gu, Yan Lu, Julian Kaiser, Martin Albrecht, Matthias Ballauff
ABSTRACT
We present a detailed study of the catalytic activity of Au/Pd nanoalloys with Au : Pd molar ratio 75 : 25 synthesized using spherical polyelectrolyte brushes (SPB) as carrier system. The reduction of 4-nitrophenol (Nip) by sodium borohydride (BH4(-)) has been used as a model reaction. This reaction proceeds in two steps: 4-nitrophenol is first reduced to 4-hydroxylaminophenol which in a second step is reduced to the final product 4-aminophenol. Both steps of the reaction proceed on the surface of the nanoparticles (Langmuir-Hinshelwood-mechanism). We use this model to analyze the experimental data obtained by catalysis with the Au/Pd-nanoalloys. Good agreements between theory and experiments were found up to 30% conversion of Nip. The kinetic parameters were compared with the data derived from neat Au and Pd nanoparticles immobilized in the same SPB carrier system. The addition of 25% molar ratio of Pd to the nanoalloys increases the reaction rate of the first step nearly 10 times compared with that of SPB-Au and 60 times compared with that of SPB-Pd. Analysis of the nanoalloy by high-resolution transmission electron microscopy suggests that the surface defects of the nanoalloys play an important role for the enhanced catalytic activity.
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, BioUltra, for molecular biology, ≥99.0% (AT)
Sigma-Aldrich
Sodium borohydride, granular, 99.99% trace metals basis
Sigma-Aldrich
Sodium borohydride, ReagentPlus®, 99%
Sigma-Aldrich
Sodium borohydride, purum p.a., ≥96% (gas-volumetric)
Sigma-Aldrich
Sodium borohydride, caplets (18 × 10 × 8 mm), 98%
Sigma-Aldrich
Sodium borohydride, powder, ≥98.0%
Sigma-Aldrich
Sodium borohydride, granular, 10-40 mesh, 98%
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, ≥98%
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, for molecular biology, ≥99%
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, BioXtra, ≥99%
Sigma-Aldrich
4-Nitrophenol solution, 10 mM
Sigma-Aldrich
VenPure® SF, powder
Sigma-Aldrich
Sodium borohydride solution, 2.0 M in triethylene glycol dimethyl ether
Sigma-Aldrich
Sodium borohydride solution, ~12 wt. % in 14 M NaOH
Sigma-Aldrich
Styrene, ReagentPlus®, contains 4-tert-butylcatechol as stabilizer, ≥99%
Sigma-Aldrich
4-Nitrophenol, ReagentPlus®, ≥99%