Merck
  • Extraction, characterization of a Ginseng fruits polysaccharide and its immune modulating activities in rats with Lewis lung carcinoma.
Extraction, characterization of a Ginseng fruits polysaccharide and its immune modulating activities in rats with Lewis lung carcinoma.
Carbohydrate polymers (2015-05-13)
Yingyu Wang, Miaoyi Huang, Ruilin Sun, Lei Pan
ABSTRACT
In this study, one polysaccharide (GFP1), with an average molecular weight of 1.4 × 10(5)Da, was isolated from Ginseng fruits. GFP1 was composed of galactose, glucose, rhamnose, and arabinose in a molar ratio of 6.1:2.0:1.1:3.2, and had a backbone mainly consisting of (1 → 6)-linked-Galp, (1 → 3,6)-linked-Galp and (1 → 3,6)-linked-Glcp residues, which was terminated with terminal (1 →)-linked-Araf or -Rhap attached to O-3 position of (1 → 3,6)-linked-Galp and (1 → 3,6)-linked-Glcp. We also evaluated the effect of GFP1 on anti-tumor immune response in Lewis lung carcinoma (LLC)-bearing mouse model and explored the possible mechanism. GPF1 could significantly inhibit tumor growth and lung metastasis in vivo, increase the relative spleen and thymus weight, promote ConA or LPS-induced spleen lymphocytes proliferation, elevate the activities of NK cell in spleen, and increase the serum concentration of interleukin-2 (IL-2) and interferon-γ (IFN-γ), as well as the ratio of CD4(+)/CD8(+) in LLC-bearing mice. All these findings implied that GFP1 could effectively inhibit tumor growth and lung metastasis via activating immune function and provide insights into the mechanism of GFP1 in the prevention and treatment of lung cancer.
Sigma-Aldrich
D-(+)-Mannose, BioUltra, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
D-(+)-Galactose, meets analytical specification of Ph. Eur., BP
Sigma-Aldrich
D-Glucuronic acid, ≥98% (GC)
Sigma-Aldrich
D-(+)-Galactose, BioXtra, ≥99% (HPLC)
Sigma-Aldrich
D-(+)-Galactose, ≥98% (HPLC)
Sigma-Aldrich
D-(+)-Galactose, ≥99% (HPLC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
D-(+)-Galactose, ≥99% (HPLC)
Sigma-Aldrich
D-(−)-Arabinose, ≥98% (GC)
Sigma-Aldrich
L-(−)-Fucose, ≥98% (GC)
Sigma-Aldrich
D-(+)-Mannose, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
D-(+)-Mannose, ≥99% (GC), wood
Sigma-Aldrich
D-(+)-Mannose, synthetic, ≥99% (GC)
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
D-(+)-Xylose, BioUltra, ≥99.0% (sum of enantiomers, HPLC)
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
D-(+)-Xylose, ≥99%
Sigma-Aldrich
D-(+)-Xylose, ≥99% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
D-(+)-Xylose, ≥99% (GC), BioXtra
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
L-Rhamnose, natural sourced, 99%, FG