Merck
miR-25 promotes glioma cell proliferation by targeting CDKN1C.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie (2015-05-12)
Jihong Zhang, Xuhai Gong, Kaiyu Tian, Dongkai Chen, Jiahang Sun, Guangzhi Wang, Mian Guo
ABSTRACT
MicroRNAs (miRNA) have oncogenic or tumor-suppressive roles in the development and growth of human glioma. Glioma development is also associated with alteration in the activities and expression of cell cycle regulators, and miRNAs are emerging as important regulators of cell cycle progression. Here, we show that miR-25 is overexpressed in 91% of examined human glioma tissues and 4 out of 6 human glioma cell lines. MiR-25 increases cell proliferation in two independent glioma cell lines. Ectopic expression of miR-25 was found to reduce CDKN1C protein levels by directly targeting its 3'-untranslated region (UTR). Notably, ablation of endogenous miR-25 rescued CDKN1C expression and significantly decreased glioma cell proliferation by facilitating normal cell cycle progression. Our clinical investigation found CDKN1C and miR-25 levels were inversely correlated. Lastly, downregulation of CDKN1 by siRNA blocked the activity of miR-25 on promoting glioma cell proliferation. Overall, our results for the first time show an oncogenic role of miR-25 in human glioma by targeting CDKN1C and that miR-25 could potentially be a therapeutic target for glioma intervention.
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
Propidium iodide solution
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Cdkn1c
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri