Merck
  • Regulation of HIF-1 alpha by the proprotein convertases furin and PC7 in human squamous carcinoma cells.
Regulation of HIF-1 alpha by the proprotein convertases furin and PC7 in human squamous carcinoma cells.
Molecular carcinogenesis (2014-01-18)
Jian Fu, Jirong Zhang, Yulan Gong, Courtney Lyons Testa, Andres J Klein-Szanto
ABSTRACT
Proprotein convertases (PC), a family of serine proteases, process cancer-related substrates such as growth factors, growth factor receptors, cell adhesion molecules, metalloproteinases, etc. HIF-1α is a major transcription factor involved in tumorigenesis by sensing intratumoral hypoxia. Furin (PCSK3) is one of the numerous target genes regulated by HIF-1α transactivation and its distribution into endosomal compartments and onto the cell surface can be triggered by hypoxia via HIF-1α. siRNAs to knockdown PCs were transfected into cells alone or in combination with different drug treatments. Protein and RNA expression levels were analyzed by Western blotting or RT-PCR, respectively. PC7 (PCSK7) and furin siRNAs upregulated HIF-1α protein under normoxic condition to a level similar to that obtained by cobalt chloride treatment, eventually leading to activation of VEGF-A synthesis in two human head and neck squamous cell carcinoma cell lines. The unchanged levels of HIF-1α mRNA expression under siRNA treatment and the additive HIF-1α induction of PC siRNAs and either cobalt chloride or the 26S ribosome inhibitor, MG-132, suggested a post-transcriptional PC-mediated regulation. Furthermore, cycloheximide chase showed that PC7/furin siRNA regulation occurred at the level of HIF-1α translation. A specific IGF-1R signaling inhibitor was able to attenuate the PC siRNA induction of HIF-1α, suggesting the involvement of the IGF-1R pathway. Thus, the data show that PCs regulate HIF-1α. Furin and PC7 siRNAs induced HIF-1α protein by increasing its translation, resulting in upregulation of VEGF-A. This finding may provide insight into intricate PC functions that seem to be independent from their substrate-processing activity.
Sigma-Aldrich
Cobalt(II) chloride, AnhydroBeads, −10 mesh, 99.995% trace metals basis
Sigma-Aldrich
Cycloheximide, from microbial, ≥94% (TLC)
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Pcsk6
Sigma-Aldrich
Picropodophyllotoxin, ≥96% (HPLC), powder
Sigma-Aldrich
Cycloheximide, Biotechnology Performance Certified
Sigma-Aldrich
Cycloheximide, ≥90% (HPLC)
Sigma-Aldrich
Cobalt(II) chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Cobalt(II) chloride hexahydrate, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
MISSION® esiRNA, targeting human PCSK6 (2)
Sigma-Aldrich
MISSION® esiRNA, targeting human PCSK7
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Pcsk7
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Furin
Sigma-Aldrich
Cycloheximide solution, Ready-Made Solution, microbial, 100 mg/mL in DMSO, Suitable for cell culture
Sigma-Aldrich
Acetyl chloride solution, 1 M in methylene chloride
Sigma-Aldrich
Acetyl chloride, reagent grade, 98%
Sigma-Aldrich
Acetyl chloride, reagent grade, 98%