Merck
  • Analysis of underivatized cellodextrin oligosaccharides by capillary electrophoresis with direct photochemically induced UV-detection.
Analysis of underivatized cellodextrin oligosaccharides by capillary electrophoresis with direct photochemically induced UV-detection.
Electrophoresis (2015-03-31)
Elodie Alinat, Selma Jemmali, Nathalie Delaunay, Xavier Archer, Pierre Gareil
ABSTRACT
This work focuses on the development of a CE method allowing, for the first time, the simultaneous separation of the underivatized first seven cellodextrin oligomers (glucose, cellobiose, cellotriose, cellotetraose, cellopentaose, cellohexaose, and celloheptaose), with a view to analyze the hydrolysates obtained after partial acid depolymerization of nitrocellulose, and eight carbohydrates (ribose, xylose, fructose, mannose, galactose, maltose, lactose, and sucrose), which might be potential interfering compounds in explosives samples. Separation was achieved with a highly alkaline BGE containing sodium chloride and direct mid-UV-absorbance detection was performed after photo-oxidation in the detection window. EOF was reversed to speed up the analysis using a dynamic capillary coating by hexadimethrine bromide. A central composite design was carried out to determine the effects of BGE conductivity and sodium hydroxide concentration on resolutions between neighboring peaks, and analysis time. A desirability analysis on modeled responses was applied to maximize resolutions and to minimize analysis time. The simultaneous analysis in 20 min total runtime of the 15 carbohydrates plus internal reference (naphthalene sulfonate) was carried out at 25°C with a BGE composed of 77.4 mM NaOH and 183 mM NaCl to adjust the conductivity at the optimum value. Finally, the resolution robustness was checked. This new method should also be of interest to monitor food and nonfood crop products.
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
D-Glucose-12C6, 16O6, 99.9 atom % 16O, 99.9 atom % 12C
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
D-(−)-Ribose, ≥98%
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
D-(−)-Ribose, 98%
Sigma-Aldrich
D-(+)-Maltose monohydrate, BioUltra, ≥99.0% (HPLC)
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
D-(+)-Galactose, meets analytical specification of Ph. Eur., BP
Sigma-Aldrich
D-(−)-Fructose, BioUltra, ≥99.0% (HPLC)
Sigma-Aldrich
D-(+)-Mannose, BioUltra, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
D-(−)-Fructose, ≥99% (HPLC)
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Sigma-Aldrich
D-(+)-Galactose, ≥99% (HPLC)
Sigma-Aldrich
D-(−)-Ribose, ≥99% (GC)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
D-(+)-Galactose, ≥99% (HPLC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
D-(+)-Galactose, ≥98% (HPLC)
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)