Merck
  • Interplay between chemotaxis and contact inhibition of locomotion determines exploratory cell migration.
Interplay between chemotaxis and contact inhibition of locomotion determines exploratory cell migration.
Nature communications (2015-04-09)
Benjamin Lin, Taofei Yin, Yi I Wu, Takanari Inoue, Andre Levchenko
ABSTRACT
Directed cell migration in native environments is influenced by multiple migratory cues. These cues may include simultaneously occurring attractive soluble growth factor gradients and repulsive effects arising from cell-cell contact, termed contact inhibition of locomotion (CIL). How single cells reconcile potentially conflicting cues remains poorly understood. Here we show that a dynamic crosstalk between epidermal growth factor (EGF)-mediated chemotaxis and CIL guides metastatic breast cancer cell motility, whereby cells become progressively insensitive to CIL in a chemotactic input-dependent manner. This balance is determined via integration of protrusion-enhancing signalling from EGF gradients and protrusion-suppressing signalling induced by CIL, mediated in part through EphB. Our results further suggest that EphB and EGF signalling inputs control protrusion formation by converging onto regulation of phosphatidylinositol 3-kinase (PI3K). We propose that this intricate interplay may enhance the spread of loose cell ensembles in pathophysiological conditions such as cancer, and possibly other physiological settings.
Product Description
SAFC
BIS-TRIS
Sigma-Aldrich
BIS-TRIS, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
BIS-TRIS, ≥98.0% (titration)
Sigma-Aldrich
BIS-TRIS, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, ≥98.0%
Sigma-Aldrich
BIS-TRIS, BioXtra, ≥98.0% (titration)
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Ephb3
SAFC
BIS-TRIS
Sigma-Aldrich
Goat Anti-Human IgG Antibody, Fc, FITC conjugate, 2 mg/mL, Chemicon®
Sigma-Aldrich
MISSION® esiRNA, targeting human EPHB3
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%