Merck
  • Influence of immunoprotection on genetic variability of cysteine proteinases from Haemonchus contortus adult worms.
Influence of immunoprotection on genetic variability of cysteine proteinases from Haemonchus contortus adult worms.
International journal for parasitology (2015-08-05)
S Martín, J M Molina, Y I Hernández, O Ferrer, Ma C Muñoz, A López, L Ortega, A Ruiz
ABSTRACT
The limitations associated with the use of anthelmintic drugs in the control of gastrotintestinal nematodosis, such as the emergence of anthelmintic resistance, have stimulated the study of the immunological control of many parasites. In the case of Haemonchus contortus, several vaccination trials using native and recombinant antigens have been conducted. A group of antigens with demonstrated immunoprotective value are cathepsin B - like proteolytic enzymes of the cysteine proteinase type. These enzymes, which have been observed in both excretory-secretory products and somatic extracts of H. contortus, may vary among different geographic isolates and on strains isolated from different hosts, or even from the same host, as has been demonstrated in some comparative studies of genetic variability. In the present study, we evaluated the genetic variability of the worms that fully developed their endogenous cycle in immunised sheep and goat in order to identify the alleles of most immunoprotective value. To address these objectives, groups of sheep and goats were immunised with PBS soluble fractions enriched for cysteine proteinases from adult worms of H. contortus from either a strain of H. contortus isolated from goats of Gran Canaria Island (SP) or a strain isolated from sheep of North America (NA). The results confirmed the immunoprophylactic value of this type of enzyme against haemonchosis in both sheep and goats in association with increased levels of specific IgG. The genetic analysis demonstrated that the immunisation had a genetic selection on proteinase-encoding genes. In all the immunised animals, allelic frequencies were statistically different from those observed in non-immunised control animals in the four analysed genes. The reduction in the allelic frequencies suggests that parasites expressing these proteases are selectively targeted by the vaccine, and hence they should be considered in any subunit vaccine approach to control haemonchosis in small ruminants.
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Sodium carbonate, anhydrous, powder, 99.999% trace metals basis
Sigma-Aldrich
Formaldehyde solution, for molecular biology, 36.5-38% in H2O
Sigma-Aldrich
Sodium carbonate, BioXtra, ≥99.0%
Sigma-Aldrich
Formamide, BioReagent, ≥99.5% (GC), for molecular biology
Supelco
Formaldehyde solution, stabilized with methanol, ~37 wt. % in H2O, certified reference material
Sigma-Aldrich
Formamide, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
Sodium carbonate, BioUltra, anhydrous, ≥99.5% (calc. on dry substance, T)
Sigma-Aldrich
Formaldehyde solution, for molecular biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
SAFC
Formaldehyde solution, contains 10-15% methanol as stabilizer, 37 wt. % in H2O
Sigma-Aldrich
Sodium carbonate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.5%
Sigma-Aldrich
Formamide, spectrophotometric grade, ≥99%
Sigma-Aldrich
Sodium carbonate, ACS reagent, anhydrous, ≥99.5%, powder or granules
Sigma-Aldrich
Sodium carbonate, ACS reagent (primary standard), anhydrous, 99.95-100.05% dry basis
Sigma-Aldrich
Sodium carbonate, powder, ≥99.5%, ACS reagent
Sigma-Aldrich
Sodium carbonate, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Formamide, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, powder
Sigma-Aldrich
Acrylamide, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Acrylamide, suitable for electrophoresis, ≥99% (HPLC), powder
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate