Merck
  • A Mechanochromic Luminescent Dye Exhibiting On/Off Switching by Crystalline-Amorphous Transitions.
A Mechanochromic Luminescent Dye Exhibiting On/Off Switching by Crystalline-Amorphous Transitions.
Chemistry, an Asian journal (2015-06-23)
Hiroaki Imoto, Kohei Kizaki, Kensuke Naka
ABSTRACT
A mechanochromic luminescent dye based on a simple aminomaleimide skeleton was readily synthesized in a one-pot process. It exhibited an on/off mechanochromic luminescent switching property dependent on external stimuli, unlike a traditional mechanochromic color change. The green emission was turned on by grinding in a mortar and turned off by heating or treatment with dichloromethane. In the crystalline state, two molecules were stacked by cofacial π-π interactions, which caused concentration self-quenching. The crystalline-to-amorphous transition induced by grinding removed cofacial π-π stacking, which led to intensive emission. Crystallizing processes recovered the cofacial π-π stacking, resulting in elimination of the emission. Theoretical calculations and X-ray diffraction analyses revealed that the dye molecule was distorted in the crystalline state; thus even a mechanical stimulus caused the crystalline-to-amorphous transition.
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Methanol solution, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Dichloromethane, suitable for HPLC, ≥99.9%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Dichloromethane, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Dimethyl acetylenedicarboxylate, 95%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Acetone, ≥99%, meets FCC analytical specifications
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
3-(Trifluoromethyl)aniline, ≥99%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol