Merck
  • Small interfering RNA delivery by polyethylenimine-functionalised porous silicon nanoparticles.
Small interfering RNA delivery by polyethylenimine-functionalised porous silicon nanoparticles.
Biomaterials science (2015-09-08)
M Hasanzadeh Kafshgari, M Alnakhli, B Delalat, S Apostolou, F J Harding, E Mäkilä, J J Salonen, B J Kuss, N H Voelcker
ABSTRACT
In this study, thermally hydrocarbonised porous silicon nanoparticles (THCpSiNPs) capped with polyethylenimine (PEI) were fabricated, and their potential for small interfering RNA (siRNA) delivery was investigated in an in vitro glioblastoma model. PEI coating following siRNA loading enhanced the sustained release of siRNA, and suppressed burst release effects. The positively-charged surface improved the internalisation of the nanoparticles across the cell membrane. THCpSiNP-mediated siRNA delivery reduced mRNA expression of the MRP1 gene, linked to the resistence of glioblastoma to chemotherapy, by 63% and reduced MRP1-protein levels by 70%. MRP1 siRNA loaded nanoparticles did not induce cytotoxicity in glioblastoma cells, but markedly reduced cell proliferation. In summary, the results demonstrated that non-cytotoxic cationic THCpSiNPs are promising vehicles for therapeutic siRNA delivery.
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Abcc1
Sigma-Aldrich
D.E.R. 332, used as embedding medium
Sigma-Aldrich
Osmium tetroxide, Sealed ampule.
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Os EnCat® 40, extent of labeling: 0.3 mmol/g Os loading
Sigma-Aldrich
Osmium tetroxide, ACS reagent, ≥98.0%
Sigma-Aldrich
2,4,6-Tris(dimethylaminomethyl)phenol, 95%
Sigma-Aldrich
Hydrofluoric acid, ACS reagent, 48%
Sigma-Aldrich
Osmium tetroxide, ReagentPlus®, 99.8%
Sigma-Aldrich
Hydrofluoric acid, 48 wt. % in H2O, ≥99.99% trace metals basis
Sigma-Aldrich
Epoxy embedding medium, accelerator, ≥95% (NT)
SAFC
L-Glutamine
Sigma-Aldrich
Propidium iodide solution
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
Sucrose, ≥99.5% (GC), Grade II, suitable for plant cell culture
Sigma-Aldrich
Sucrose, Grade I, ≥99% (GC), suitable for plant cell culture
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Dodecenylsuccinic anhydride, mixture of isomers, technical grade, 90%
Sigma-Aldrich
Osmium tetroxide solution, suitable for electron microscopy, 4% in H2O
Sigma-Aldrich
Osmium tetroxide solution, suitable for electron microscopy, 2% in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Sucrose, BioUltra, for molecular biology, ≥99.5% (HPLC)
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
MISSION® esiRNA, targeting human ABCC1
Sigma-Aldrich
L-Glutamine
Sigma-Aldrich
Sucrose, for molecular biology, ≥99.5% (GC)