Merck
  • A selective biomarker for confirming nitrofurazone residues in crab and shrimp using ultra-performance liquid chromatography-tandem mass spectrometry.
A selective biomarker for confirming nitrofurazone residues in crab and shrimp using ultra-performance liquid chromatography-tandem mass spectrometry.
Analytical and bioanalytical chemistry (2015-10-03)
Shuai Zhang, Yuanming Guo, Zhongyong Yan, Xiumei Sun, Xiaojun Zhang
ABSTRACT
Reliably detecting nitrofurazone (NFZ) residues in farmed crab and shrimp was previously hindered by lack of appropriately specific analytical methodology. Parent NFZ rapidly breaks down in meat, and the commonly used side-chain metabolite, semicarbazide (SEM), is non-specific as it occurs naturally in crustacean shell often leading to 'false positive' detections in meat. Using 5-nitro-2-furaldehyde (NF) as marker metabolite, following pre-column derivatization with 2,4-dinitrophenylhydrazine (DNPH), ultra-performance liquid chromatography-tandem mass spectrometry (UPLC-MS/MS) analysis in negative electrospray ionization mode enabled confirmation of NFZ residues in deliberately treated whole crab, crab meat and shrimp meat, with a limit of detection (LOD) and limit of quantification (LOQ) below 1 ng g(-1). Meanwhile, the derivatives of DNPH-NF were synthesized for the first time, purified by preparative liquid chromatography and structure characterized with nuclear magnetic resonance spectroscopy ((1)H-NMR). The purity of derivative was checked by ultra-performance liquid chromatography-tunable ultraviolet (UPLC-TUV), and the contents were beyond 99.9%. For comparison purposes, crustacean samples were analysed using both NF and SEM marker metabolites. NFZ treatment was revealed by both NF and SEM marker metabolites, but untreated crab also showed measurable levels of SEM which could potentially be misinterpreted as evidence of illegal NFZ use.
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in acetic acid
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in diethyl ether
Sigma-Aldrich
Ammonium acetate, ACS reagent, ≥97%
Sigma-Aldrich
Hydrochloric acid, 37 wt. % in H2O, 99.999% trace metals basis
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Hydrogen chloride solution, 2.0 M in diethyl ether
Sigma-Aldrich
Hydrogen chloride solution, 4.0 M in dioxane
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Sulfuric acid, ACS reagent, 95.0-98.0%
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Ammonium acetate, BioXtra, ≥98%
Sigma-Aldrich
Ammonium acetate solution, for molecular biology, 7.5 M
Sigma-Aldrich
Ammonium acetate, for molecular biology, ≥98%
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Ammonium acetate, reagent grade, ≥98%
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
Sulfuric acid, 99.999%
Sigma-Aldrich
Ammonium acetate, 99.999% trace metals basis
Sigma-Aldrich
5-Nitro-2-furaldehyde, 99%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Ammonium acetate, ≥99.99% trace metals basis