Merck
  • In vitro study of normoxic epidermal growth factor receptor-induced hypoxia-inducible factor-1-alpha, vascular endothelial growth factor, and BNIP3 expression in head and neck squamous cell carcinoma cell lines: Implications for anti-epidermal growth factor receptor therapy.
In vitro study of normoxic epidermal growth factor receptor-induced hypoxia-inducible factor-1-alpha, vascular endothelial growth factor, and BNIP3 expression in head and neck squamous cell carcinoma cell lines: Implications for anti-epidermal growth factor receptor therapy.
Head & neck (2014-05-07)
Pablo Secades, Inés Saenz de Santa-María, Anna Merlo, Carlos Suarez, María-Dolores Chiara
ABSTRACT
We previously showed that activation of epidermal growth factor receptor (EGFR) induces hypoxia inducible factor-1α (HIF-1α) in head and neck squamous cell carcinoma (HNSCC) cells. In this study, we have furthered this by investigating the mechanism of HIF-1α activation by epidermal growth factor (EGF) and its association with the sensitivity to gefitinib. EGFR/HIF-1α signaling was tested by immunoblot, polymerase chain reaction (PCR), cell proliferation, and apoptosis assays. HIF-1α accumulated in cells overexpressing EGF and phosphorylated epidermal growth factor receptor (pEGFR), phosphatidylinositol-3-kinase (pPI3K), and mitogen-activated protein kinase (pMAPK). EGF-induced expression of HIF-1α and its targets, vascular endothelial growth factor (VEGF) and BNIP3, were blocked by gefitinib and PI3K-inhibitors and MAPK-inhibitors. HIF-1α-siRNAs abrogated EGF-induced BNIP3 but not VEGF expression. Gefitinib inhibited cell proliferation and induced apoptosis more strongly in cells with constitutively active EGFR/HIF-1α signaling than in cells lacking activation of these pathways. HIF-1α-siRNA treatment reduced sensitivity to gefitinib. The search for molecular predictors of sensitivity to gefitinib in HNSCC should be extended to the activation status of EGFR-downstream pathways, phosphorylated protein kinase B, pMAPK, and HIF-1α.
Sigma-Aldrich
HEPES, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
Carbon dioxide, ≥99.8%
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
SAFC
HEPES
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Dand5
SAFC
HEPES
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
Carbon-12C dioxide, 99.99 atom % 12C
Sigma-Aldrich
MISSION® esiRNA, targeting human SP1
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
Carbon-12C dioxide, 99.9 atom % 12C
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Sp1