Merck
  • 111In-cetuximab as a diagnostic agent by accessible epidermal growth factor (EGF) receptor targeting in human metastatic colorectal carcinoma.
111In-cetuximab as a diagnostic agent by accessible epidermal growth factor (EGF) receptor targeting in human metastatic colorectal carcinoma.
Oncotarget (2015-06-13)
Ying-Hsia Shih, Cheng-Liang Peng, Shin-Yu Lee, Ping-Fang Chiang, Cheng-Jung Yao, Wuu-Jyh Lin, Tsai-Yueh Luo, Ming-Jium Shieh
ABSTRACT
Colorectal adenocarcinoma is a common cause death cancer in the whole world. The aim of this study is to define the 111In-cetuximab as a diagnosis tracer of human colorectal adenocarcinoma. In this research, cell uptake, nano-SPECT/CT scintigraphy, autoradiography, biodistribution and immunohitochemical staining of EGF receptor were included. HCT-116 and HT-29 cell expressed a relatively high and moderate level of EGF receptor, respectively. The nano-SPECT/CT image of 111In-cetuximab showed tumor radiation uptake of subcutaneous HCT-116 xenograft tumor was higher than SW-620. Autoradiography image also showed that tumor of HCT-116 had high 111In-cetuximab uptake. Mice that bearing CT-26 in situ xenograft colorectal tumors showed similar high uptake in vivo and ex vivo through nano-SPECT/CT imaging at 72 hours. Metastatic HCT-116/Luc tumors demonstrated the highest uptake at 72 hours after the injection of 111In-cetuximab. Relatively, results of 111In-DTPA showed that metabolism through urinary system, especially in the kidney. The quantitative analysis of biodistribution showed count value of metastatic HCT-116/Luc tumors that treated with 111In-cetuximab had a significant difference (P < 0.05) compared with that treated with 111In-DTPA after injection 72 hours. Result of immunohistologic staining of EGF receptor also showed high EGF receptor expression and uptake in metastatic colorectal tumors. In summary, we suggested that 111In-cetuximab will be a potential tool for detecting EGF receptor expression in human metastatic colorectal carcinoma.
Sigma-Aldrich
EGF human, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
hEGF, EGF, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
MISSION® esiRNA, targeting human EGF
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
Epidermal Growth Factor, human, animal component free, EGF, recombinant, expressed in Escherichia coli, >97% (SDS-PAGE)