Merck
Temperature dependence of amino acid hydrophobicities.
Proceedings of the National Academy of Sciences of the United States of America (2015-06-03)
Richard Wolfenden, Charles A Lewis, Yang Yuan, Charles W Carter
ABSTRACT
The hydrophobicities of the 20 common amino acids are reflected in their tendencies to appear in interior positions in globular proteins and in deeply buried positions of membrane proteins. To determine whether these relationships might also have been valid in the warm surroundings where life may have originated, we examined the effect of temperature on the hydrophobicities of the amino acids as measured by the equilibrium constants for transfer of their side-chains from neutral solution to cyclohexane (K(w > c)). The hydrophobicities of most amino acids were found to increase with increasing temperature. Because that effect is more pronounced for the more polar amino acids, the numerical range of K(w > c) values decreases with increasing temperature. There are also modest changes in the ordering of the more polar amino acids. However, those changes are such that they would have tended to minimize the otherwise disruptive effects of a changing thermal environment on the evolution of protein structure. Earlier, the genetic code was found to be organized in such a way that--with a single exception (threonine)--the side-chain dichotomy polar/nonpolar matches the nucleic acid base dichotomy purine/pyrimidine at the second position of each coding triplet at 25 °C. That dichotomy is preserved at 100 °C. The accessible surface areas of amino acid side-chains in folded proteins are moderately correlated with hydrophobicity, but when free energies of vapor-to-cyclohexane transfer (corresponding to size) are taken into consideration, a closer relationship becomes apparent.
Sigma-Aldrich
Carbon, mesoporous, less than 100 ppm Al, Ti, Fe, Ni, Cu, and Zn combined
Sigma-Aldrich
Carbon, mesoporous, nanopowder, less than 500 ppm Al, Ti, Fe, Ni, Cu, and Zn combined
Sigma-Aldrich
Carbon, mesoporous, nanopowder, graphitized, less than 250 ppm Al, Ti, Fe, Ni, Cu, and Zn combined
Sigma-Aldrich
Carbon, mesoporous, hydrophilic pore surface
Sigma-Aldrich
Carbon, mesoporous
Sigma-Aldrich
2-Methylpropane, 99%
Sigma-Aldrich
Methane-12C, 13C-depleted, 99.99 atom % 12C
Sigma-Aldrich
Carbon nanofibers, graphitized (iron-free), composed of conical platelets, D × L 100 nm × 20-200 μm
Carbon - Vitreous, foil, 10x10mm, thickness 4.0mm, glassy carbon
Carbon - Vitreous, rod, 50mm, diameter 1.0mm, glassy carbon
Carbon - Vitreous, foil, 100x100mm, thickness 6.0mm, glassy carbon
Carbon - Vitreous, rod, 5mm, diameter 3.0mm, glassy carbon
Carbon - Vitreous, rod, 200mm, diameter 3.0mm, glassy carbon
Carbon - Vitreous, rod, 200mm, diameter 5.0mm, glassy carbon
Carbon - Vitreous, foam, 150x150mm, 0.05g.cmué, porosity 96.5%, 24 pores/cm
Carbon - Vitreous, foam, 150x150mm, thickness 2.5mm, bulk density 0.05g/cm3, porosity 96.5%
Carbon - Vitreous, rod, 100mm, diameter 7.0mm, glassy carbon
Carbon - Vitreous, foil, 10x10mm, thickness 1.0mm, glassy carbon
Carbon - Vitreous, foam, 150x150mm, thickness 3.2mm, bulk density 0.05g/cm3, porosity 96.5%
Carbon - Vitreous, rod, 100mm, diameter 1.0mm, glassy carbon
Carbon - Vitreous, foil, 8x8mm, thickness 0.5mm, glassy carbon
Carbon - Vitreous, foil, 100x100mm, thickness 1.0mm, glassy carbon
Carbon - Vitreous, foil, 25x25mm, thickness 0.5mm, glassy carbon
Carbon - Vitreous, foil, 50x50mm, thickness 4.0mm, glassy carbon
Carbon - Vitreous, rod, 100mm, diameter 3.0mm, glassy carbon
Carbon - Vitreous, foam, 300x300mm, thickness 20mm, bulk density 0.05g/cm3, porosity 96.5%
Carbon - Vitreous, tube, 100mm, outside diameter 10mm, inside diameter 3mm, wall thickness 3.5mm, glassy carbon
Carbon - Vitreous, foam, 300x300mm, thickness 30mm, bulk density 0.05g/cm3, porosity 96.5%
Carbon - Vitreous, tube, 50mm, outside diameter 10mm, inside diameter 3mm, wall thickness 3.5mm, glassy carbon
Carbon - Vitreous, foil, 50x50mm, thickness 1.0mm, glassy carbon