Merck
  • Synthesis of Amphiphilic Ru(II) Heteroleptic Complexes Based on Benzo[1,2-b:4,5-b']dithiophene: Relevance of the Half-Sandwich Complex Intermediate and Solvent Compatibility.
Synthesis of Amphiphilic Ru(II) Heteroleptic Complexes Based on Benzo[1,2-b:4,5-b']dithiophene: Relevance of the Half-Sandwich Complex Intermediate and Solvent Compatibility.
Chemistry (Weinheim an der Bergstrasse, Germany) (2015-09-12)
Maxence Urbani, María Medel, Sangeeta Amit Kumar, Mine Ince, Ashok N Bhaskarwar, David González-Rodríguez, Michael Grätzel, Mohammad Khaja Nazeeruddin, Tomás Torres
ABSTRACT
The detailed synthesis and characterization of four ruthenium(II) complexes [RuLL'(NCS)2 ] is reported, in which L represents a 2,2'-bipyridine ligand functionalized at the 4,4' positions with benzo[1,2-b:4,5-b']dithiophene derivatives (BDT) and L' is 2,2'-bipyridine-4,4'-dicarboxylic acid unit (dcbpy) (NCS=isothiocyanate). The reaction conditions were adapted and optimized for the preparation of these amphiphilic complexes with a strong lipophilic character. The photovoltaic performances of these complexes were tested in TiO2 dye-sensitized solar cell (DSSC) achieving efficiencies in the range of 3-4.5 % under simulated one sun illumination (AM1.5G).
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
n-Butyllithium solution, 1.6 M in hexanes
Sigma-Aldrich
2,2′-Bipyridine-4,4′-dicarboxylic acid, 98%
Sigma-Aldrich
Methanol solution, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 20% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 10% in H2O
Sigma-Aldrich
Toluene, anhydrous, 99.8%
Sigma-Aldrich
n-Butyllithium solution, 11.0 M in hexanes
Sigma-Aldrich
n-Butyllithium solution, 2.0 M in cyclohexane
Sigma-Aldrich
n-Butyllithium solution, 2.5 M in hexanes
Sigma-Aldrich
n-Butyllithium solution, 2.7 M in heptane
Sigma-Aldrich
Hexane, Laboratory Reagent, ≥95%
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Sigma-Aldrich
Hexane, ReagentPlus®, ≥99%
Sigma-Aldrich
Ammonium thiocyanate, powder
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Supelco
Ammonium thiocyanate, 99.99% trace metals basis
Sigma-Aldrich
Ammonium thiocyanate, ACS reagent, ≥97.5%
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Sigma-Aldrich
Lithium diisopropylamide solution, 1.0 M in THF/hexanes
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Methanol
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Lithium diisopropylamide, 97%
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Triethylamine, ≥99.5%
Sigma-Aldrich
Triethylamine, ≥99%
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)