Merck
Association of vitamin D3 with alveolar bone regeneration in dogs.
Journal of cellular and molecular medicine (2015-03-11)
Hsiang-Hsi Hong, Tzung-Hai Yen, Adrienne Hong, Ting-An Chou
ABSTRACT
Designed sockets prepared on the mandibles of nine Beagle dogs were divided into three groups: Calcitriol +Alloplast, Alloplast and Empty. Five of the nine dogs received Vit.D3 and calcium supplement (Vit.D/Ca group), while the other four dogs without supplements were assigned to Non-Vit.D/Ca group. After 4 weeks, the extent of vertical ridge resorption (VRR), bone density (density), new bone formation (NBF) and implant stability quotient (ISQ) were measured. Following systemic Vit.D/Ca administration, the Empty subgroup showed significant differences from the Calcitriol + Alloplast subgroup on variants NBF/Density/VRR and the Alloplast subgroup on items NBF/Density/ISQ/VRR. Alternatively, the Calcitriol + Alloplast subgroup revealed higher values of NBF/Density/ISQ (P < 0.001) and a lower VRR value (P = 0.001) than the Alloplast subgroup. Although there were no significant differences in NBF (P = 0.349), density (P = 0.796), ISQ (P = 0.577) and VRR (0.979) comparisons on alloplast treatment between the Vit.D/Ca and Non-Vit.D/Ca groups, local application with Calcitriol + Alloplast demonstrated better NBF/Density/ISQ (P = 0.02 to <0.001) effects than which of Alloplast subgroups. Consequently, the results showed that both systemic and local vitamin D3 treatment might accelerate bone regeneration in dogs. Within the using dose, systemic vitamin D3 treatment displayed a superior stimulating effect than local vitamin D3 application did.
Sigma-Aldrich
Hydroxyapatite, reagent grade, powder, synthetic
Sigma-Aldrich
Hydroxyapatite, synthetic, 99.8% trace metals basis (excludes Mg)
Sigma-Aldrich
Hydroxyapatite, purum p.a., ≥90% (as Ca3(PO4)2, KT)
Sigma-Aldrich
Hydroxyapatite, puriss., meets analytical specification of Ph. Eur., BP, FCC, E341, ≥90% (calculated on glowed substance)
Sigma-Aldrich
Hydroxyapatite, nanopowder, <200 nm particle size (BET), ≥97%, synthetic
Sigma-Aldrich
Calcium phosphate tribasic, 34.0-40.0% Ca basis
Sigma-Aldrich
Hydroxyapatite, Type I, buffered aqueous suspension
Sigma-Aldrich
Calcium phosphate tribasic, suitable for plant cell culture, BioReagent, powder
Sigma-Aldrich
Hydroxyapatite, nanopowder, <200 nm particle size (BET), contains 5 wt. % silica as dopant, synthetic
Sigma-Aldrich
Hydroxyapatite, nanoparticles, dispersion, 10 wt. % in H2O, <200 nm particle size (BET)
Sigma-Aldrich
1α,25-Dihydroxyvitamin D2 solution, 50 μg/mL in ethanol, 95% (CP)
Sigma-Aldrich
1α,25-Dihydroxyvitamin D2 solution, 100 μg/mL in ethanol, 95% (CP)
Sigma-Aldrich
1α,25-Dihydroxyvitamin D3, ≥99% (HPLC)
Supelco
1α,25-Dihydroxyvitamin D2 solution, 5 μg/mL in ethanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
1α,25-Dihydroxyvitamin D3, ≥97.0% (HPLC)