Merck
  • Patterned Plasmonic Nanoparticle Arrays for Microfluidic and Multiplexed Biological Assays.
Patterned Plasmonic Nanoparticle Arrays for Microfluidic and Multiplexed Biological Assays.
Analytical chemistry (2015-10-24)
Jie He, Michelle Boegli, Ian Bruzas, William Lum, Laura Sagle
ABSTRACT
For applications ranging from medical diagnostics and drug screening to chemical and biological warfare detection, inexpensive, rapid-readout, portable devices are required. Localized surface plasmon resonance (LSPR) technologies show substantial promise toward meeting these goals, but the generation of portable, multiplexed and/or microfluidic devices incorporating sensitive nanoparticle arrays is only in its infancy. Herein, we have combined photolithography with Hole Mask Colloidal lithography to pattern uniform nanoparticle arrays for both microfluidic and multiplexed devices. The first proof-of-concept study is carried out with 5- and 7-channel microfluidic devices to acquire one-shot binding curves and protein binding kinetic data. The second proof-of-concept study involved the fabrication of a 96-spot plate that can be inserted into a standard plate reader for the multiplexed detection of protein binding. This versatile fabrication technique should prove useful in next generation chips for bioassays and genetic screening.
Sigma-Aldrich
11-Mercaptoundecanoic acid, 98%
Sigma-Aldrich
11-Mercaptoundecanoic acid, 95%
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
Hydrofluoric acid, 48 wt. % in H2O, ≥99.99% trace metals basis
Sigma-Aldrich
Hydrofluoric acid, ACS reagent, 48%
Sigma-Aldrich
Ammonium fluoride, ACS reagent, ≥98.0%
Sigma-Aldrich
Ammonium fluoride, ≥99.99% trace metals basis
Sigma-Aldrich
N,O-Bis(trimethylsilyl)acetamide, synthesis grade, ≥95%
Sigma-Aldrich
1-Octanethiol, ≥98.5%
Sigma-Aldrich
Trichloro(octadecyl)silane, ≥90%
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Glycerol, FCC, FG
Sigma-Aldrich
Glycerol, for molecular biology, ≥99.0%
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Glycerol, ACS reagent, ≥99.5%
Sigma-Aldrich
Glycerol, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Glycerol, BioUltra, for molecular biology, anhydrous, ≥99.5% (GC)