Merck
  • Copper acutely impairs behavioral function and muscle acetylcholinesterase activity in zebrafish (Danio rerio).
Copper acutely impairs behavioral function and muscle acetylcholinesterase activity in zebrafish (Danio rerio).
Ecotoxicology and environmental safety (2015-09-20)
Gabriela M B Haverroth, Chariane Welang, Riciéri N Mocelin, Daniela Postay, Kanandra T Bertoncello, Francini Franscescon, Denis B Rosemberg, Jacir Dal Magro, Cristiane L Dalla Corte
ABSTRACT
Copper is a heavy metal found at relatively high concentrations in surface waters around the world. Copper is a micronutrient at low concentrations and is essential to several organisms. At higher concentrations copper can become toxic, which reveal the importance of studying the toxic effects of this metal on the aquatic life. Thus, the objective of this study was to evaluate the toxic effects of copper on the behavior and biochemical parameters of zebrafish (Danio rerio). Zebrafish were exposed for 24h at a concentration of 0.006 mg/L Cu. After the exposure period, behavioral profile of animals was recorded through 6 min using two different apparatuses tests: the Novel Tank and the Light-Dark test. After behavioral testing, animals were euthanized with a solution of 250 mg/L of tricaine (MS-222). Brain, muscle, liver and gills were extracted for analysis of parameters related to oxidative stress and accumulation of copper in these tissues. Acetylcholinesterase (AChE) activity was determined in brain and muscle. Results showed acute exposure to copper induces significant changes in behavioral profile of zebrafish by changing locomotion and natural tendency to avoid brightly lit area. On the other hand, there were no significant effects on parameters related to oxidative stress. AChE activity decreased significantly in zebrafish muscle, but there were no significant changes in cerebral AChE activity. Copper levels in tissues did not increase significantly compared to the controls. Taken together, these results indicate that a low concentration of copper can acutely affect behavioral profile of adult zebrafish which could be partially related to an inhibition on muscle AChE activity. These results reinforce the need of additional tests to establishment of safe copper concentrations to aquatic organisms and the importance of behavioral parameters in ecotoxicological studies.
Sigma-Aldrich
Copper(II) sulfate pentahydrate, 99.999% trace metals basis
Sigma-Aldrich
5,5′-Dithiobis(2-nitrobenzoic acid), ReagentPlus®, 99%
Sigma-Aldrich
Copper(II) sulfate pentahydrate, 99.995% trace metals basis
Sigma-Aldrich
Copper(II) sulfate pentahydrate, suitable for plant cell culture, ≥98%
Sigma-Aldrich
Copper(II) sulfate pentahydrate, BioReagent, suitable for cell culture, ≥98%
Sigma-Aldrich
5,5′-Dithiobis(2-nitrobenzoic acid), ≥98%, BioReagent, suitable for determination of sulfhydryl groups
Sigma-Aldrich
2-Thiobarbituric acid, ≥98%
Sigma-Aldrich
Copper(II) sulfate pentahydrate, ACS reagent, ≥98.0%
Sigma-Aldrich
Copper(II) sulfate pentahydrate, ReagentPlus®, ≥98.0%
Sigma-Aldrich
1,1,3,3-Tetramethoxypropane, 99%
Sigma-Aldrich
Acetylthiocholine iodide, ≥98% (TLC), powder or crystals
Sigma-Aldrich
Acetylthiocholine iodide, ≥99.0% (AT)