Merck
  • Immunogenicity of GX301 cancer vaccine: Four (telomerase peptides) are better than one.
Immunogenicity of GX301 cancer vaccine: Four (telomerase peptides) are better than one.
Human vaccines & immunotherapeutics (2015-02-26)
Daniela Fenoglio, Alessia Parodi, Rosa Lavieri, Francesca Kalli, Francesca Ferrera, Augusto Tagliamacco, Andrea Guastalla, Maria Giuseppina Lamperti, Mauro Giacomini, Gilberto Filaci
ABSTRACT
Peptide540-548, peptide611-626, peptide672-686 and peptide766-780, which are derived from human telomerase, constitute the immunogenic component of the GX301 cancer vaccine. The relative immunogenicity of these peptides is unknown, thus it is unsure whether their combined use offers real advantages over single peptide stimulation. Hence, this study compared the number of specific immune responses and responders to each peptide, as well as to their mixture (meaning the co-presence of the 4 peptides in the same culture well), achieved after ex vivo stimulation of PBMC from 21, HLA-A2+ (n.11) or HLA-A2- (n.10), healthy donors. The study was performed on freshly collected PBMC (T0) and on PBMC stimulated for 10 d with single peptides or their mixture (T1). Peptide-specific immune responses were analyzed by Elispot and cytokine intracellular staining by flow cytometry. The results showed that each peptide induced specific immune responses in some subjects, with different panels of responders among the peptides. Moreover, the numbers of responses and responders to the single peptides or their mixture were comparable. Importantly, the overall number of responders to the 4 peptides was higher than to each single peptide, or to their mixture, both at T0 and T1. These data demonstrate the immunogenicity of each of the 4 GX301 telomerase peptides. Moreover, they show the advantage of multi-peptide over single peptide stimulation, providing a clear support to their combined administration in vaccination protocols. However, the data pose a warning against peptide administration as a mixture due to possible interference phenomena during antigen presentation processes.
Sigma-Aldrich
Interleukin-2 human, IL-2, recombinant, expressed in HEK 293 cells, suitable for cell culture, endotoxin tested
Sigma-Aldrich
Interleukin-2, human, Animal-component free, recombinant, expressed in E. coli, suitable for cell culture
Sigma-Aldrich
3,3′-Diiodo-L-thyronine (T2) hydrochloride, 98% (CP)
Sigma-Aldrich
IL-7 from rat, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Sigma-Aldrich
IL-2 from rat, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Sigma-Aldrich
Interleukin-2 human, recombinant, expressed in E. coli, ~10000 U/mL
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Interleukin-2 from mouse, IL-2, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
IL-7 human, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC)
Sigma-Aldrich
Interleukin-2 from mouse, IL-2, recombinant, expressed in E. coli, carrier free
Sigma-Aldrich
Interleukin-2 human, recombinant, expressed in Pichia pastoris, suitable for cell culture
Sigma-Aldrich
Interleukin-2 human, IL-2, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Sigma-Aldrich
Interleukin-7 from mouse, ≥97% (SDS-PAGE), recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Sigma-Aldrich
Interleukin-7 human, ≥98% (SDS-PAGE), recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture