Merck
  • Eucalyptus urograndis stem proteome is responsive to short-term cold stress.
Eucalyptus urograndis stem proteome is responsive to short-term cold stress.
Genetics and molecular biology (2015-08-15)
Gabriela de Almeida Leonardi, Natália Aparecida Carlos, Paulo Mazzafera, Tiago Santana Balbuena
ABSTRACT
Eucalyptus urograndis is a hybrid eucalyptus of major economic importance to the Brazilian pulp and paper industry. Although widely used in forest nurseries around the country, little is known about the biochemical changes imposed by environmental stress in this species. In this study, we evaluated the changes in the stem proteome after short-term stimulation by exposure to low temperature. Using two-dimensional gel electrophoresis coupled to high-resolution mass spectrometry-based protein identification, 12 proteins were found to be differentially regulated and successfully identified after stringent database searches against a protein database from a closely related species (Eucalyptus grandis). The identification of these proteins indicated that the E. urograndis stem proteome responded quickly to low temperature, mostly by down-regulating specific proteins involved in energy metabolism, protein synthesis and signaling. The results of this study represent the first step in understanding the molecular and biochemical responses of E. urograndis to thermal stress.
Sigma-Aldrich
Urea-12C, 99.9 atom % 12C
Sigma-Aldrich
3-[(3-Cholamidopropyl)dimethylammonio]-1-propanesulfonate hydrate, 98%
Sigma-Aldrich
Thiourea, ACS reagent, ≥99.0%
Sigma-Aldrich
Urea solution, 40 % (w/v) in H2O
Sigma-Aldrich
Thiourea, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Urea solution, BioUltra, ~8 M in H2O
Supelco
Urea, 8 M (after reconstitution with 16 mL high purity water)
Sigma-Aldrich
Phenol, natural, 97%, FG
Sigma-Aldrich
Cinnamyl alcohol, 98%
Sigma-Aldrich
Bromophenol Blue, ACS reagent
Sigma-Aldrich
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (GC)
Sigma-Aldrich
Phenol, ≥96.0% (calc. on dry substance, T)
Sigma-Aldrich
Phenol, puriss. p.a., ACS reagent, reag. Ph. Eur., 99.0-100.5%
Sigma-Aldrich
Phenol, BioUltra, for molecular biology, TE-saturated, ~73% (T)
Sigma-Aldrich
Urea, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)
Sigma-Aldrich
Phenol, unstabilized, purified by redistillation, ≥99%
Sigma-Aldrich
Phenol, unstabilized, ReagentPlus®, ≥99%
Sigma-Aldrich
Phenol, contains hypophosphorous as stabilizer, loose crystals, ACS reagent, ≥99.0%
Sigma-Aldrich
Urea, BioUltra, for molecular biology, 99% (T)
Sigma-Aldrich
Phenol, ≥99%
Sigma-Aldrich
Cinnamyl alcohol, ≥98%, FG
Sigma-Aldrich
Phenol solution, BioReagent, Saturated with 0.01 M citrate buffer, pH 4.3 ± 0.2, for molecular biology
Sigma-Aldrich
Liquified Phenol, ≥89.0%
Sigma-Aldrich
Phenol solution, BioReagent, Equilibrated with 10 mM Tris HCl, pH 8.0, 1 mM EDTA, for molecular biology
Sigma-Aldrich
Urea, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Sigma-Aldrich
Urea, ReagentPlus®, ≥99.5%, pellets
Sigma-Aldrich
Urea, powder, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Urea, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Phenol, for molecular biology
Sigma-Aldrich
Bromophenol Blue, titration: suitable