Merck
  • Functional characterization of a yellow laccase from Leucoagaricus gongylophorus.
Functional characterization of a yellow laccase from Leucoagaricus gongylophorus.
SpringerPlus (2015-11-07)
Priscila Tomie Leme Ike, Ariele C Moreira, Fernando G de Almeida, Douglas Ferreira, Willian Garcia Birolli, Andre Luiz Meleiro Porto, Dulce Helena F Souza
ABSTRACT
In this work we have identified, using mass spectrometry, two laccases produced by Leucoagaricus gongylophorus. One of them, Lac1Lg, was isolated, purified and characterized. Lac1Lg, a monomeric enzyme, was studied using ABTS and syringaldazine substrates. Lac1Lg presented kcat/Km almost threefold higher for syringaldazine than for ABTS, showing a higher catalytic efficiency of Lac1Lg for syringaldazine. The interference of several metal ions and substances in the laccase activity were evaluated. Lac1Lg did not absorb at 600 nm, which is a characteristic of so-called yellow laccases. Lac1Lg also was able to oxidize non-phenolic substrate (anthracene) in the absence of an exogenous mediator, showing that the enzyme has potential to explore in biotechnological processes. Our Lac1Lg three-dimensional molecular model, constructed using homology modeling, showed that the Lac1Lg catalytic site is very closed to blue laccases.
Sigma-Aldrich
Hydroxyapatite, nanopowder, <200 nm particle size (BET), contains 5 wt. % silica as dopant, synthetic
Sigma-Aldrich
Hydroxyapatite, nanoparticles, dispersion, 10 wt. % in H2O, <200 nm particle size (BET)
Sigma-Aldrich
Hydroxyapatite, purum p.a., ≥90% (as Ca3(PO4)2, KT)
Sigma-Aldrich
Hydroxyapatite, Type I, buffered aqueous suspension
Sigma-Aldrich
Calcium phosphate tribasic, suitable for plant cell culture, BioReagent, powder
Sigma-Aldrich
Syringaldazine, indicator for laccase and peroxidase activity
Sigma-Aldrich
Hydroxyapatite, puriss., meets analytical specification of Ph. Eur., BP, FCC, E341, ≥90% (calculated on glowed substance)
Sigma-Aldrich
Hydroxyapatite, reagent grade, powder, synthetic
Sigma-Aldrich
Syringaldazine, 98%
Sigma-Aldrich
Hydroxyapatite, synthetic, 99.8% trace metals basis (excludes Mg)
Sigma-Aldrich
Calcium phosphate tribasic, 34.0-40.0% Ca basis
Sigma-Aldrich
Hydroxyapatite, nanopowder, <200 nm particle size (BET), ≥97%, synthetic
Sigma-Aldrich
2,2′-Azino-bis(3-ethylbenzothiazoline-6-sulfonic acid) diammonium salt, ≥98% (HPLC)
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
2,2′-Azino-bis(3-ethylbenzothiazoline-6-sulfonic acid) diammonium salt, tablet, 10 mg substrate per tablet
Sigma-Aldrich
Calcium chloride, anhydrous, powder, 99.99% trace metals basis
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.99% trace metals basis
Sigma-Aldrich
Calcium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Sodium phosphate, 96%
Sigma-Aldrich
Calcium chloride, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Sigma-Aldrich
2,2′-Azino-bis(3-ethylbenzothiazoline-6-sulfonic acid) diammonium salt, ≥98% (HPLC)
Sigma-Aldrich
Calcium chloride