Merck
  • Biochemical Characterization of the DASH-Type Cryptochrome CryD From Fusarium fujikuroi.
Biochemical Characterization of the DASH-Type Cryptochrome CryD From Fusarium fujikuroi.
Photochemistry and photobiology (2015-07-29)
Marta Castrillo, Adrian Bernhardt, Javier Ávalos, Alfred Batschauer, Richard Pokorny
ABSTRACT
Proteins from the cryptochrome/photolyase family utilize UV-A, blue or even red light to achieve such diverse functions as repair of DNA lesions by photolyases and signaling by cryptochromes. DASH-type cryptochromes retained the ability to repair cyclobutane pyrimidine dimers (CPDs) in single-stranded DNA regions in vitro. However, most organisms possess conventional CPD photolyases responsible for repair of these lesions in vivo. Recent work showed that the DASH-type cryptochrome CryD plays a regulatory role in diverse light-dependent processes in Fusarium fujikuroi. Here, we report our in vitro studies on heterologously expressed FfCryD. The purified protein contains N(5) ,N(10) -methenyltetrahydrofolate and flavin adenine dinucleotide as cofactors. Photoreduction and DNA photorepair experiments confirmed that FfCryD is active in light-driven electron transfer processes. However, the protein showed comparable affinities for CPD-comprising and undamaged DNA probes. Surprisingly, after purification, full-length FfCryD as well as a truncated version containing only the PHR domain bound RNA which influenced their behavior in vitro. Moreover, binding of FfCryD to RNA indicates a putative role in RNA metabolism or in posttranscriptional control of gene expression.
Sigma-Aldrich
Riboflavin 5′-phosphate sodium salt hydrate, meets USP testing specifications
Sigma-Aldrich
Riboflavin 5′-monophosphate sodium salt hydrate, ≥95% (HPLC)
Sigma-Aldrich
Ampicillin, anhydrous, 96.0-102.0% (anhydrous basis)
Sigma-Aldrich
Ampicillin, meets USP testing specifications
Sigma-Aldrich
Riboflavin 5′-monophosphate sodium salt hydrate, synthetic, ≥70% (HPLC)
Sigma-Aldrich
Riboflavin 5′-monophosphate sodium salt hydrate, 73-79% (fluorimetric)
Sigma-Aldrich
Riboflavin 5′-monophosphate sodium salt hydrate, suitable for electrophoresis, suitable for acrylamide photopolymerization, ≥70%, powder