Merck
  • The Chilean wild raspberry (Rubus geoides Sm.) increases intracellular GSH content and protects against H2O2 and methylglyoxal-induced damage in AGS cells.
The Chilean wild raspberry (Rubus geoides Sm.) increases intracellular GSH content and protects against H2O2 and methylglyoxal-induced damage in AGS cells.
Food chemistry (2015-10-17)
Felipe Jiménez-Aspee, Cristina Theoduloz, Felipe Ávila, Samanta Thomas-Valdés, Claudia Mardones, Dietrich von Baer, Guillermo Schmeda-Hirschmann
ABSTRACT
The Chilean raspberry Rubus geoides Sm. (Rosaceae) is a native species occurring in the Patagonia. Five R. geoides samples were assessed for phenolic content and composition, antioxidant activity, effect on total reduced glutathione (GSH) synthesis and protective effect against H2O2 and methylglyoxal (MGO)-induced stress in epithelial gastric AGS cells. The HPLC-DAD/ESI-MS profiles allowed the tentative identification of 39 phenolics including flavonol glycosides and tannins. R. geoides presented higher total phenolic and flavonoid content than Rubus idaeus. Two out of the five phenolic enriched R. geoides extracts (PEEs) exhibited better antioxidant activity than R. idaeus in the DPPH, FRAP and TEAC assays. A significant cytoprotective activity was observed when AGS cells were pre-incubated with extracts and subsequently challenged with H2O2 or MGO. Treatment with the PEEs increased the intracellular GSH content. R. geoides fruit extracts may induce the activation of intracellular protection mechanisms against oxidative and dicarbonyl-induced stress.
Sigma-Aldrich
Potassium persulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
Supelco
Phenol solution, 5000 μg/mL in methanol, certified reference material
Supelco
Phenol solution, certified reference material, 500 μg/mL in methanol
Sigma-Aldrich
Liquified Phenol, ≥89.0%
Sigma-Aldrich
Methylglyoxal solution, ~40% in H2O
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, ≥97.5%
Sigma-Aldrich
Phenol, for molecular biology
Sigma-Aldrich
Phenol, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
Phenol solution, BioReagent, Saturated with 0.01 M citrate buffer, pH 4.3 ± 0.2, for molecular biology
Sigma-Aldrich
Phenol solution, BioReagent, Equilibrated with 10 mM Tris HCl, pH 8.0, 1 mM EDTA, for molecular biology
Sigma-Aldrich
Phenol, BioUltra, for molecular biology, TE-saturated, ~73% (T)
Sigma-Aldrich
Phenol, puriss. p.a., ACS reagent, reag. Ph. Eur., 99.0-100.5%
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, purum, ≥99.0% (TLC)
Sigma-Aldrich
Phenol, ≥96.0% (calc. on dry substance, T)
Sigma-Aldrich
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (GC)
Sigma-Aldrich
Phenol, puriss., ≥99.5% (GC), meets analytical specification of Ph. Eur., BP, USP, crystalline (detached)
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Potassium persulfate, 99.99% trace metals basis
Sigma-Aldrich
Sodium carbonate-12C, 99.9 atom % 12C
Sigma-Aldrich
Phenol, ≥99%
Sigma-Aldrich
1,1-Diphenyl-2-picrylhydrazine, 97%
Sigma-Aldrich
Phenol, contains hypophosphorous as stabilizer, loose crystals, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium bicarbonate-12C, 99.9 atom % 12C
Sigma-Aldrich
Phenol, unstabilized, purified by redistillation, ≥99%
Sigma-Aldrich
Phenol, unstabilized, ReagentPlus®, ≥99%
Sigma-Aldrich
Gallic acid, 97.5-102.5% (titration)
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Phenol, natural, 97%, FG
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source