Merck
  • A polydopamine-modified optical fiber SPR biosensor using electroless-plated gold films for immunoassays.
A polydopamine-modified optical fiber SPR biosensor using electroless-plated gold films for immunoassays.
Biosensors & bioelectronics (2015-07-15)
Se Shi, Libing Wang, Rongxin Su, Boshi Liu, Renliang Huang, Wei Qi, Zhimin He
ABSTRACT
A sensitive and stable electroless-plated gold film for the preparation of an optical fiber surface plasmon resonance (SPR) sensor is presented in this work, together with a facile antibody immobilization method. Gold nanoparticles were uniformly adsorbed onto the surface of an optical fiber forming a film with a thickness of approximately 56.3 nm. The sensor had a high sensitivity with 2054 nm/RIU and 3980 nm/RIU in the refractive index ranges of 1.333-1.359 and 1.359-1.386, respectively. An SPR biosensor was developed based on polydopamine-modified gold film (PDA-Au), which was fabricated by a simple and quick spontaneous polymerization of dopamine (DA) on the gold film. When goat anti-human IgG antibodies were immobilized, the PDA-Au surface had a larger resonant wavelength shift of 66.21 nm compared with the traditional 11-mercaptoundecanoic acid-modified gold film (MUA-Au) surface. In addition, the PDA-Au surface enabled the sensitive and selective determination of human IgG down to a concentration of 2 μg mL(-1) with a high sensitivity of 0.41 nm per μg mL(-1). The PDA-Au surface exhibited an approximately four fold higher sensitivity and an about seven fold lower LOD than the MUA-Au surface to human IgG.
Sigma-Aldrich
11-Mercaptoundecanoic acid, 98%
Sigma-Aldrich
Hydroxylamine hydrochloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium borohydride solution, ~12 wt. % in 14 M NaOH
Sigma-Aldrich
Sodium borohydride solution, 2.0 M in triethylene glycol dimethyl ether
Sigma-Aldrich
11-Mercaptoundecanoic acid, 95%
Sigma-Aldrich
Dopamine hydrochloride
Sigma-Aldrich
VenPure® SF, powder
Sigma-Aldrich
Hydroxylamine hydrochloride, 99.995% trace metals basis
Sigma-Aldrich
Potassium phosphate monobasic, powder, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Potassium phosphate monobasic, for molecular biology, ≥98.0%
Sigma-Aldrich
Potassium phosphate monobasic, ReagentPlus®
Sigma-Aldrich
Sodium borohydride, powder, ≥98.0%
Sigma-Aldrich
Hydroxylamine solution, 50 wt. % in H2O
Sigma-Aldrich
Sodium borohydride, ReagentPlus®, 99%
Sigma-Aldrich
Hydroxylamine solution, 50 wt. % in H2O, 99.999%
Sigma-Aldrich
Potassium phosphate monobasic, BioUltra, for molecular biology, anhydrous, ≥99.5% (T)
Sigma-Aldrich
(3-Aminopropyl)trimethoxysilane, 97%
Sigma-Aldrich
Sodium borohydride, granular, 99.99% trace metals basis
Sigma-Aldrich
N-Hydroxysuccinimide, 98%
Sigma-Aldrich
Ethanolamine hydrochloride, anhydrous, free-flowing, Redi-Dri, ≥99.0%
Sigma-Aldrich
Sodium borohydride, purum p.a., ≥96% (gas-volumetric)
Sigma-Aldrich
Potassium phosphate monobasic, 99.99% trace metals basis
Sigma-Aldrich
Sodium borohydride, granular, 10-40 mesh, 98%
Sigma-Aldrich
Sodium borohydride, caplets (18 × 10 × 8 mm), 98%
Sigma-Aldrich
Ethanolamine hydrochloride, ≥99.0%
Sigma-Aldrich
1,2-Dichloroethane, anhydrous, 99.8%