Merck
  • Stress-Induced Glucocorticoid Release Upregulates Uncoupling Protein-2 Expression and Enhances Resistance to Endotoxin-Induced Lethality.
Stress-Induced Glucocorticoid Release Upregulates Uncoupling Protein-2 Expression and Enhances Resistance to Endotoxin-Induced Lethality.
Neuroimmunomodulation (2015-06-16)
Emiko Kasahara, Atsuo Sekiyama, Mika Hori, Daisuke Kuratsune, Naoki Fujisawa, Dai Chida, Keiichi Hiramoto, Jiawei Li, Haruki Okamura, Masayasu Inoue, Seiichi Kitagawa
ABSTRACT
Although psychological and/or physiological stress has been well documented to influence immune responses, the precise mechanism for immunomodulation remains to be elucidated. The present work describes the role of the hypothalamic-pituitary-adrenal (HPA) axis in the mechanism of stress-mediated enhanced-resistance to lethality after lipopolysaccharide (LPS) injection. Preconditioning with restraint stress (RS) resulted in enhanced activation of the HPA axis in response to LPS injection and suppressed LPS-induced release of proinflammatory cytokines and nitric oxide metabolites. Melanocortin 2 receptor-deficient mice (MC2R(-/-)) failed to increase plasma levels of glucocorticoids in response to LPS injection, and exhibited high sensitivity to LPS-induced lethality with enhanced release of proinflammatory cytokines as compared with MC2R(+/-) mice. Real-time PCR analysis revealed that RS induced upregulation of uncoupling protein-2 (UCP2) in macrophages in the lung and the liver of MC2R(+/-), but not of MC2R(-/-), mice. In addition, RS increased UCP2-dependent uncoupling activity of isolated mitochondria from the liver of MC2R(+/-), but not of MC2R(-/-), mice. In vitro study revealed that corticosterone and dexamethasone directly increased UCP2 expression in mouse RAW 264.7 macrophages and suppressed the generation of LPS-induced mitochondrial reactive oxygen species (ROS) and TNF-α production. Knockdown of UCP2 by small interfering RNA blunted the dexamethasone action for suppressing LPS-induced mitochondrial ROS and TNF-α production. The present work suggests that RS enhances activation of the HPA axis to release glucocorticoids and upregulation of UCP2 in macrophages, thereby increasing the resistance to endotoxin-induced systemic inflammation and death.
Sigma-Aldrich
Sodium deoxycholate, BioXtra, ≥98.0% (dry matter, NT)
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
Sodium deoxycholate, ≥97% (titration)
Sigma-Aldrich
Mifepristone, ≥98%
SAFC
Sodium deoxycholate
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Corticosterone, ≥98.5% (HPLC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Ucp2
Sigma-Aldrich
MISSION® esiRNA, targeting human UCP2
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Corticosterone, ≥92%
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri