Merck
  • Lactate dehydrogenase B is associated with the response to neoadjuvant chemotherapy in oral squamous cell carcinoma.
Lactate dehydrogenase B is associated with the response to neoadjuvant chemotherapy in oral squamous cell carcinoma.
PloS one (2015-05-15)
Wenyi Sun, Xiaomin Zhang, Xu Ding, Huaiqi Li, Meiyu Geng, Zuoquan Xie, Heming Wu, Min Huang
ABSTRACT
Oral squamous cell carcinoma (OSCC) comprises a subset of head and neck squamous cell carcinoma (HNSCC) with poor therapeutic outcomes and high glycolytic dependency. Neoadjuvant chemotherapy regimens of docetaxel, cisplatin and 5-fluorouracil (TPF) are currently accepted as standard regimens for HNSCC patients with a high risk of distant metastatic spread. However, the antitumor outcomes of TPF neoadjuvant chemotherapy in HNSCC remain controversial. This study investigated the role of lactate dehydrogenase B (LDHB), a key glycolytic enzyme catalyzing the inter-conversion between pyruvate and lactate, in determining chemotherapy response and prognosis in OSCC patients. We discovered that a high protein level of LDHB in OSCC patients was associated with a poor response to TPF regimen chemotherapy as well as poor overall survival and disease-free survival. Our in-depth study revealed that high LDHB expression conferred resistance to taxol but not 5-fluorouracil or cisplatin. LDHB deletion sensitized OSCC cell lines to taxol, whereas the introduction of LDHB decreased sensitivity to taxol treatment. Taxol induced a pronounced impact on LDHB-down-regulated OSCC cells in terms of apoptosis, G2/M phase cell cycle arrest and energy metabolism. In conclusion, our study highlighted the critical role of LDHB in OSCC and proposed that LDHB could be used as a biomarker for the stratification of patients for TPF neoadjuvant chemotherapy and the determination of prognosis in OSCC patients.
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Propidium iodide solution
Sigma-Aldrich
MISSION® esiRNA, targeting human LDHA
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Ldha