Merck
  • Molecular engineering of a dual emission near-infrared ratiometric fluorophore for the detection of pH at the organism level.
Molecular engineering of a dual emission near-infrared ratiometric fluorophore for the detection of pH at the organism level.
The Analyst (2015-05-29)
Bo-Lin Wang, Chuang Jiang, Kun Li, Yan-Hong Liu, Yongmei Xie, Xiao-Qi Yu
ABSTRACT
A near-infrared ratiometric fluorophore (NIR-HBT) was rationally designed and constructed by expanding both the excitation and emission wavelength of the classical ratiometric fluorophore 2-(benzothiazol-2-yl)phenol (HBT) into the near-infrared region. The NIR-HBT was easily synthesized by incorporating the HBT module into the hemicyanine skeleton and showed evident NIR ratiometric fluorophore characteristics. Further application of the new fluorophore for pH detection demonstrated that NIR-HBT possesses superior overall analytical performance and NIR-HBT was successfully applied for detection of acidosis caused by inflammation in living animal tissue, which indicated the potential application value of NIR-HBT in biological imaging and sensing.
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 0.05 % (v/v) TMS
Sigma-Aldrich
(Tyr[SO3H]27)Cholecystokinin fragment 26-33 Amide, ≥97% (HPLC), powder
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 0.1 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, 99.8 atom % D
Sigma-Aldrich
Chloroform-d, "100%", 99.95 atom % D
Sigma-Aldrich
Chloroform-d, ≥99.8 atom % D, anhydrous
Sigma-Aldrich
Chloroform-d, ≥99.8 atom % D, contains 0.5 wt. % silver foil as stabilizer, 0.03 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, "100%", 99.96 atom % D, contains 0.5 wt. % silver wire as stabilizer
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, ≥99.8 atom % D, contains 0.5 wt. % silver foil as stabilizer
Sigma-Aldrich
Chloroform-d, "100%", 99.96 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 1 % (v/v) TMS